Skip to Content
Merck
All Photos(1)

Key Documents

113239

Sigma-Aldrich

1-Phenyldodecane

97%

Synonym(s):

Dodecylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5(CH2)11CH3
CAS Number:
Molecular Weight:
246.43
Beilstein:
1909107
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.482 (lit.)

bp

185-188 °C/15 mmHg (lit.)
331 °C (lit.)

mp

3 °C (lit.)

density

0.856 g/mL at 25 °C (lit.)

functional group

phenyl

SMILES string

CCCCCCCCCCCCc1ccccc1

InChI

1S/C18H30/c1-2-3-4-5-6-7-8-9-10-12-15-18-16-13-11-14-17-18/h11,13-14,16-17H,2-10,12,15H2,1H3

InChI key

KWKXNDCHNDYVRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Phenyldodecane acts as a pseudo-stationary phase (PSP) marker in the preparation of microemulsion.

Biochem/physiol Actions

1-Phenyldodecane was degraded to phenylacetic acid by an oil-degrading bacterium.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Shaoqiang Hu et al.
Journal of chromatography. A, 1216(45), 7932-7940 (2009-09-29)
A novel procedure for in situ assembling a complex chiral selector, di-n-butyl l-tartrate-boric acid complex, by the reaction of di-n-butyl l-tartrate with boric acid in a running buffer was reported and its application in the enantioseparation of beta-blockers and structural
Studies of the carcinogenesis and tumorigenesis of skin applications of dodecylbenzene on hairless mice.
C B Assogne
British journal of industrial medicine, 47(5), 356-358 (1990-05-01)
J Campos-García et al.
Applied and environmental microbiology, 65(8), 3730-3734 (1999-07-31)
Pseudomonas aeruginosa W51D is able to grow by using branched-chain dodecylbenzene sulfonates (B-DBS) or the terpenic alcohol citronellol as a sole source of carbon. A mutant derived from this strain (W51M1) is unable to degrade citronellol but still grows on
Ding-xi Cheng et al.
Guang pu xue yu guang pu fen xi = Guang pu, 27(1), 104-107 (2007-03-30)
There is a self-polymerization equilibrium system between the cation of the fluorescent dye-safranine T and its dimer. In the presence of appropriate amounts of anionic surfactant SDBS, the above mentioned equilibrium can be adjusted by a quantitative addition of protein
C Ruffo et al.
Ecotoxicology and environmental safety, 8(3), 275-279 (1984-06-01)
Pyridine and nitrilotriacetic acid sodium salt as models of soluble compounds and linear dodecylbenzene, branched dodecylbenzene, and stearic acid as models of insoluble compounds were compared for their biodegradability in three tests: the ODCE modified test and the Sturm and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service