Skip to Content
Merck
All Photos(3)

Key Documents

434965

Sigma-Aldrich

2-Hydroxyethyl cellulose

average Mv ~90,000

Synonym(s):

Cellulose hydroxyethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23
Pricing and availability is not currently available.

form

powder

Quality Level

autoignition temp.

725 °F

mol wt

average Mv ~90,000

extent of labeling

2.5 mol per mol cellulose (M.S.)

viscosity

100-180 cP(lit.)

density

0.6 g/mL at 25 °C (lit.)

SMILES string

O1C(C(C(C(C1CO)OC)O)OCCO)OC2C(OC(C(C2O)O)OC4C(OC(C(C4O)O)C)CO)COC3OC(C(C(C3O)O)OC)CO

InChI

1S/C29H52O21/c1-10-15(34)16(35)24(13(8-33)45-10)49-28-20(39)18(37)25(50-29-26(43-5-4-30)21(40)23(42-3)12(7-32)47-29)14(48-28)9-44-27-19(38)17(36)22(41-2)11(6-31)46-27/h10-40H,4-9H2,1-3H3

InChI key

CWSZBVAUYPTXTG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Hydroxyethyl cellulose (HEC) is a water soluble biopolymer that is majorly used as a thickener and an adhesive. It has water retention properties and good film forming ability for protective covering.[1][2]
Non-ionic water soluble polymer. Aqueous solutions are pseudoplastic. Readily disperses without lumping.

Application

Thickener, protective colloid, binder, stabilizer and suspending agent.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical modification of cellulose extracted from sugarcane bagasse: Preparation of hydroxyethyl cellulose
Abdel-Halim ES
Arabian Journal of Chemistry, 7(3), 362-371 (2014)
Distribution of substituents in O-(2-hydroxyethyl) cellulose
Lindberg B, et al.
Carbohydrate Research, 170(2), 207-214 (1987)
Mingyun Sun et al.
Electrophoresis, 32(22), 3233-3240 (2011-10-20)
Double-stranded (ds) DNA fragments over a wide size range were successfully separated in blended polymer matrices by microfluidic chip electrophoresis. Novel blended polymer matrices composed of two types of polymers with three different molar masses were developed to provide improved
Salomé dos Santos et al.
Journal of colloid and interface science, 383(1), 63-74 (2012-07-17)
Associative aqueous mixtures over a range of concentrations of double- (ds) or single- (ss) stranded DNA with dilute or semidilute solutions of two cationic derivatives of hydroxyethyl cellulose (cat-HEC and cat-HMHEC,(1) the latter carrying grafted hydrophobic groups), were studied. The
Barbra A Richardson et al.
Journal of acquired immune deficiency syndromes (1999), 63(1), 120-125 (2013-01-22)
To compare the 2 control arms of HPTN 035 [a hydroxyethylcellulose (HEC) gel control arm and a no-gel control arm] to assess the behavioral effects associated with gel use and direct causal effects of the HEC gel on sexually transmitted

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service