Skip to Content
Merck
All Photos(2)

Key Documents

435015

Sigma-Aldrich

Methyl 2-hydroxyethyl cellulose

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

extent of labeling

0.06-0.50 mol hydroxyethyl per mol cellulose (M.S.)
8 wt. % hydroxyethyl
26 wt. % methoxy
1.3-2.2 mol methyl per mol cellulose (D.S.)

surface tension

45-55 dyn/cm, 20 °C, 0.1 wt. %

viscosity

15,000-20,500 cP, 2 wt. % in H2O(20 °C, Brookfield, spindle #6) (20 rpm)(lit.)

transition temp

flocculation range 60-90 °C (0.5 wt. %)

SMILES string

O3[C@H]([C@@H]([C@H]([C@@H]([C@H]3COC)OC)OC)OC)O[C@@H]4[C@H](O[C@H]([C@@H]([C@H]4OC)OC)OC)COC.O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)O[C@@H]2[C@H](O[C@H]([C@@H]([C@H]2O)O)O)CO.OCCO

InChI key

JTWZVKMWOCBJEV-RXUSYSPUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Water retention aid, thickening agent, protective colloid, suspending agent, binder and stabilizer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rondedrick Sinville et al.
Electrophoresis, 29(23), 4751-4760 (2008-12-05)
We have developed a new method for the analysis of low abundant point mutations in genomic DNA using a combination of an allele-specific ligase detection reaction (LDR) with free-solution conjugate electrophoresis (FSCE) to generate and analyze the genetic products. FSCE
M G Lee
Journal of clinical and hospital pharmacy, 7(1), 67-70 (1982-03-01)
A method is reported for the analysis of prostaglandin PGE2 in methylhydroxyethylcellulose gel. Stability studies using the method have shown that the degradation of PGE in the gel is a first order process at 4 degrees and 25 degrees C.
K M Picker et al.
Pharmaceutical development and technology, 3(1), 31-41 (1998-04-09)
The aim of this research was to investigate the elastic-plastic deformation behavior of the cellulose ethers hydroxypropylmethylcellulose (HPMC), hydroxyethylmethylcellulose (HEMC), and sodium carboxymethylcellulose (NaCMC) at relative humidities (RH) of 38, 57, and 75% and assess how the release of drugs
Su Jeong Ban et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50(5), 1716-1721 (2012-03-06)
The effect of hydroxyethyl methylcellulose (HEMC) with different viscosities on the glucose metabolism and antioxidative defense system in high fat-fed mice was investigated. The mice were randomly divided into five dietary groups: normal control diet (NC), high fat diet (HF)
Roland Adden et al.
Analytical chemistry, 78(4), 1146-1157 (2006-02-16)
Hydroxyethylmethyl celluloses (HEMC, DS(Me) 1.46-1.66, DS(HE) = 0.14-0.17) have been analyzed with respect to their methyl and hydroxyethyl pattern in the glucosyl units and along the polymer chain. Methyl groups were located by GLC/MS after direct hydrolysis, reduction, and acetylation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service