Skip to Content
Merck
All Photos(3)

Key Documents

434981

Sigma-Aldrich

2-Hydroxyethyl cellulose

average Mv ~1,300,000

Synonym(s):

Hydroxyethylcellulose

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder

autoignition temp.

725 °F

mol wt

average Mv ~1,300,000

extent of labeling

2.5 mol per mol cellulose (M.S.)

viscosity

3,400-5,000 cP, 1 wt. % in H2O(25 °C, Brookfield, spindle #4) (30 rpm)(lit.)

density

0.6 g/mL at 25 °C (lit.)

SMILES string

O1C(C(C(C(C1CO)OC)O)OCCO)OC2C(OC(C(C2O)O)OC4C(OC(C(C4O)O)C)CO)COC3OC(C(C(C3O)O)OC)CO

InChI

1S/C29H52O21/c1-10-15(34)16(35)24(13(8-33)45-10)49-28-20(39)18(37)25(50-29-26(43-5-4-30)21(40)23(42-3)12(7-32)47-29)14(48-28)9-44-27-19(38)17(36)22(41-2)11(6-31)46-27/h10-40H,4-9H2,1-3H3

InChI key

CWSZBVAUYPTXTG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Non-ionic water soluble polymer. Aqueous solutions are pseudoplastic. Readily disperses without lumping.

Application

Thickener, protective colloid, binder, stabilizer and suspending agent.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prem N Gupta et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 46(5), 315-322 (2012-03-01)
Mucosally-administered vaccine strategies are widely investigated as a promising means of preventing HIV infection. This study describes the development of liposomal gel formulations, and novel lyophilised variants, comprising HIV-1 envelope glycoprotein, CN54gp140, encapsulated within neutral, positively charged or negatively charged
Bernice Wright et al.
Regenerative medicine, 7(3), 295-307 (2012-05-19)
Therapeutic limbal epithelial stem cells could be managed more efficiently if clinically validated batches were transported for 'on-demand' use. In this study, corneal epithelial cell viability in calcium alginate hydrogels was examined under cell culture, ambient and chilled conditions for
Maoqiang Xue et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 912, 1-7 (2013-01-08)
Microporous anodic aluminum oxide (AAO) membranes were modified by 3-glycidoxypropyltrimethoxysilane to produce terminal epoxy groups. These were used to covalently link hydroxyethyl celluloses (HEC) to amplify reactive groups of AAO membrane. The hydroxyl groups of HEC-AAO composite membrane were further
Liangzhu Li et al.
PloS one, 7(5), e37381-e37381 (2012-05-23)
Sifuvirtide is a proven effective HIV-1 entry inhibitor and its safety profile has been established for systemic administration. The present study evaluated the potential of sifuvirtide formulated in a universal gel for topical use as a microbicide candidate for preventing
Shao-Ching Hung et al.
Journal of agricultural and food chemistry, 60(44), 11149-11156 (2012-10-23)
Cationic hydroxyethyl cellulose (cHEC) was supplemented in a high-fat diet to determine if this new soluble fiber had an effect on hypercholesterolemia and dyslipidemia associated with cardiovascular disease using Golden Syrian hamster as an animal model. Supplementation of 3-5% cHEC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service