Skip to Content
Merck
All Photos(3)

Key Documents

419273

Sigma-Aldrich

Sodium carboxymethyl cellulose

viscosity 50-200 cP , c=4% H2O at 25­°C

Synonym(s):

Carboxymethylcellulose sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder

autoignition temp.

698 °F

extent of labeling

0.7 carboxymethyl groups per anhydroglucose unit

viscosity

50-200 cP

mp

274 °C (dec.)

SMILES string

[Na].OC(C(O)C(O)C=O)C(O)CO.OC(=O)C

InChI

1S/C6H12O6.C2H4O2.Na/c7-1-3(9)5(11)6(12)4(10)2-8;1-2(3)4;/h1,3-6,8-12H,2H2;1H3,(H,3,4);

InChI key

DPXJVFZANSGRMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium carboxymethyl cellulose (CMC) is a water dispersible sodium salt of carboxy-methyl ether of cellulose that forms a clear colloidal solution. It is a hygroscopic material that has the ability to absorb more than 50% of water at high humidity. It is also a natural polymeric derivative that can be used in detergents, food and textile industries.

Application

CMC can be used to stabilize palladized iron nanoparticles, which can further be utilized in the dichlorination of contaminated subsurfaces. It may also be used as a polymeric matrix to form a composite with a crystalline nanofibril for the development of sustainable bio-based polymers. CMC can also bind with a hard carbon electrode for the fabrication of sodium ion-batteries.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sodium carboxymethyl cellulose as a potential binder for hard-carbon negative electrodes in sodium-ion batteries
Dahbi M, et al.
Electrochemical Communications, 44(1), 66-69 (2014)
Preparation and characterization of sodium carboxymethyl cellulose/cotton linter cellulose nanofibril composite films
Oun AA and Rhim J
Carbohydrate Polymers, 127(1), 101-109 (2015)
M Rahman et al.
Nature communications, 10(1), 3712-3712 (2019-08-20)
Nanopore-based single nanoparticle detection has recently emerged as a vibrant research field with numerous high-impact applications. Here, we introduce a programmable optofluidic chip for nanopore-based particle analysis: feedback-controlled selective delivery of a desired number of biomolecules and integration of optical
Radiation preparation and swelling behavior of sodium carboxymethyl cellulose hydrogels
Liu P, et al.
Radiation Physics and Chemistry, 63(3-6), 525-528 (2002)
Handbook of pharmaceutical granulation technology (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service