Skip to Content
Merck
All Photos(1)

Key Documents

339601

Sigma-Aldrich

1-Bromo-4-(trifluoromethoxy)benzene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4OCF3
CAS Number:
Molecular Weight:
241.01
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.461 (lit.)

bp

80 °C/50 mmHg (lit.)

density

1.622 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

FC(F)(F)Oc1ccc(Br)cc1

InChI

1S/C7H4BrF3O/c8-5-1-3-6(4-2-5)12-7(9,10)11/h1-4H

InChI key

SEAOBYFQWJFORM-UHFFFAOYSA-N

General description

1-Bromo-4-(trifluoromethoxy)benzene on treatment with lithium diisopropylamide (LIDA) at -100°C gives 5-bromo-2-(trifluoromethoxy)phenyllithium and at -75°C it eliminates lithium bromide, thus generating 1,2-dehydro-4-(trifluoromethoxy)benzene.

Application

1-Bromo-4-(trifluoromethoxy)benzene was used in the synthesis of 4-{[4-[(2-ethylhexyloxy)-6-(4-morpholinylmethyl)-4-trifluoromethyl][1,1-biphenyl]-3-yl]methyl}morpholine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1, 2-Didehydro-3-and-4-(trifluoromethoxy) benzene: The ?Aryne? Route to 1-and 2-(Trifluoromethoxy) naphthalenes.
Schlosser M and Castagnetti E.
European Journal of Organic Chemistry, 2001(21), 3991-3997 (2001)
Novel poly (p-phenylenevinylene) s derivatives with CF3-phenyl substituent for light-emitting diodes.
Jin Y, et al.
Bull. Korean Chem. Soc., 26(5), 795-801 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service