B59004
3-Bromobenzotrifluoride
99%
Synonym(s):
1-Bromo-3-(trifluoromethyl)benzene, 3-Bromo-α,α,α-trifluorotoluene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
BrC6H4CF3
CAS Number:
Molecular Weight:
225.01
Beilstein:
1449557
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
form
liquid
refractive index
n20/D 1.473 (lit.)
bp
151-152 °C (lit.)
density
1.613 g/mL at 25 °C (lit.)
SMILES string
FC(F)(F)c1cccc(Br)c1
InChI
1S/C7H4BrF3/c8-6-3-1-2-5(4-6)7(9,10)11/h1-4H
InChI key
NNMBNYHMJRJUBC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
109.4 °F - closed cup
Flash Point(C)
43 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service