Skip to Content
Merck
All Photos(2)

Key Documents

C26402

Sigma-Aldrich

4-Chlorobenzotrifluoride

98%

Synonym(s):

1-Chloro-4-(trifluoromethyl)benzene, 4-Chloro-α,α,α-trifluorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CF3
CAS Number:
Molecular Weight:
180.55
Beilstein:
510203
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.446 (lit.)

bp

136-138 °C (lit.)

mp

−36 °C (lit.)

density

1.353 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)c1ccc(Cl)cc1

InChI

1S/C7H4ClF3/c8-6-3-1-5(2-4-6)7(9,10)11/h1-4H

InChI key

QULYNCCPRWKEMF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Flam. Liq. 3 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

109.4 °F - closed cup

Flash Point(C)

43 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J M Yuan et al.
Toxicology and applied pharmacology, 111(1), 107-115 (1991-10-01)
Toxicokinetics of p-chloro-alpha,alpha,alpha-trifluorotoluene (CTFT) after administration as an aqueous alpha-cyclodextrin (alpha-CD) molecular encapsulation suspension (alpha-CD vehicle) or as a corn oil solution (corn oil vehicle) were compared. Male F344 rats were administered intragastrically CTFT in alpha-CD vehicle or corn oil
A Macrì et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 25(10), 781-786 (1987-10-01)
The subchronic oral toxicity of 4-chloro-alpha, alpha, alpha-trifluorotoluene (CTT) was assessed in Sprague-Dawley rats. Four groups of six male and six female rats were treated daily for 28 days, by gavage, with doses of 0, 10, 100 and 1000 mg
[Determination of the para-chlorobenzotrifluoride and toluene content in water].
V S Kozlova et al.
Gigiena i sanitariia, (1)(1), 35-37 (1986-01-01)
[Experimental study of the toxicodynamics of parachlorobenzotrifluoride].
O G Charyev et al.
Gigiena i sanitariia, (7)(7), 26-28 (1985-07-01)
Metabolism of p-chlorobenzotrifluoride by rats.
G B Quistad et al.
Journal of agricultural and food chemistry, 31(3), 585-589 (1983-05-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service