290564
4-(Trifluoromethyl)benzyl bromide
98%
Synonym(s):
α′-Bromo-α,α,α-trifluoro-p-xylene, 4-(Bromomethyl)benzotrifluoride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
98%
form
solid
refractive index
n20/D 1.484 (lit.)
bp
65-69 °C/5 mmHg (lit.)
mp
29-33 °C (lit.)
density
1.546 g/mL at 25 °C (lit.)
functional group
bromo
fluoro
SMILES string
FC(F)(F)c1ccc(CBr)cc1
InChI
1S/C8H6BrF3/c9-5-6-1-3-7(4-2-6)8(10,11)12/h1-4H,5H2
InChI key
IKSNDOVDVVPSMA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Vibrational characteristics of 4-(trifluoromethyl)benzylbromide have been investigated.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
eLife, 10 (2021-05-13)
Phospholamban (PLN) is a mini-membrane protein that directly controls the cardiac Ca2+-transport response to β-adrenergic stimulation, thus modulating cardiac output during the fight-or-flight response. In the sarcoplasmic reticulum membrane, PLN binds to the sarco(endo)plasmic reticulum Ca2+-ATPase (SERCA), keeping this enzyme's
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 116, 84-95 (2013-08-06)
In this work, the vibrational characteristics of 4-(trifluoromethyl) benzylbromide (TFMBB) have been investigated and both the experimental and theoretical vibrational data indicate the presence of functional groups in the title molecule. The density functional theoretical (DFT) computations were performed at
Journal of medicinal chemistry, 60(22), 9263-9274 (2017-10-14)
Azapeptide analogues of growth hormone releasing peptide-6 (GHRP-6) exhibit promising affinity, selectivity, and modulator activity on the cluster of differentiation 36 receptor (CD36). For example, [A
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service