Skip to Content
Merck
All Photos(1)

Key Documents

590592

Sigma-Aldrich

Hafnium(IV) chloride

purified by sublimation, 99.9% trace metals basis

Synonym(s):

Hafnium tetrachloride, Tetrachlorohafnium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HfCl4
CAS Number:
Molecular Weight:
320.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

1 mmHg ( 190 °C)

Quality Level

Assay

99.9% trace metals basis

form

powder

purified by

sublimation

impurities

≤1500.0 ppm Trace Metal Analysis

SMILES string

Cl[Hf](Cl)(Cl)Cl

InChI

1S/4ClH.Hf/h4*1H;/q;;;;+4/p-4

InChI key

PDPJQWYGJJBYLF-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Hafnium(IV) chloride can be used as a catalyst:
  • To synthesize α-aminophosphonates via the one-pot three-component reaction of aldehyde, amine, and phosphite.
  • For the conversion of carbohydrates into 5-hydroxymethylfurfural in ionic liquids.
  • For parallel synthesis of 1,2-disubstituted benzimidazoles from N-substituted phenylenediamines and aldehydes.
It can also be used as:
  • A precursor to prepare hafnium oxide nanoparticles via sol-gel method.
  • A starting material to synthesize bulky guanidinato hafnium(iv) chloridecomplexes.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Hafnium oxide nanoparticles synthesized via sol-gel route for an efficient detection of volatile organic compounds at room temperature
Anamitra Chattopadhyay and Jhasaketan Nayak
Materials Science in Semiconductor Processing, 139 , 106336-106336 (2022)
Activated carbon supported hafnium (IV) chloride as an efficient, recyclable, and facile removable catalyst for expeditious parallel synthesis of benzimidazoles
Xiao-Chong Peng, et al.
Catalysts, 10, 436-436 (2020)
Catalytic conversion of carbohydrates into 5-hydroxymethylfurfural by Hafnium (IV) chloride in ionic liquids
Zehui Zhang, et al.
Starch, 64, 770-775 (2012)
Synthesis and crystal structures of bulky guanidinato zirconium (IV) and hafnium (IV) chloride complexes
Cameron Jones, et al.
Journal of Chemical Crystallography, 42, 866-870 (2012)
Highly efficient synthesis of ?-aminophosphonates catalyzed by hafnium(IV) chloride
Xiao-Chuan Li, et al.
Tetrahedron Letters, 57, 1782-1785 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service