Merck
  • Hydroaminations of unactivated alkenes with basic alkylamines: group 4 metal halide catalysts and Brønsted-acid organocatalysts.
Hydroaminations of unactivated alkenes with basic alkylamines: group 4 metal halide catalysts and Brønsted-acid organocatalysts.
Organic & biomolecular chemistry (2007-06-07)
Lutz Ackermann, Ludwig T Kaspar, Andreas Althammer
ABSTRACT
Two distinct economical catalysts for intramolecular hydroaminations of electronically unactivated alkenes with basic amines are described, which are based on (a) group 4 metal halides under basic reaction conditions or (b) Brønsted-acid organocatalysts.
Sigma-Aldrich
Hafnium(IV) chloride, purified by sublimation, 99.9% trace metals basis
Sigma-Aldrich
Zirconium(IV) chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Hafnium(IV) chloride, 98%
Sigma-Aldrich
Zirconium(IV) chloride, ≥99.5% trace metals basis
Sigma-Aldrich
Zirconium(IV) chloride, ≥99.9% trace metals basis