241946
1-Iodopentane
98%
Synonym(s):
n-Amyl iodide, Pentyl iodide
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
CH3(CH2)4I
CAS Number:
Molecular Weight:
198.05
Beilstein:
1730982
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
form
liquid
contains
copper as stabilizer
refractive index
n20/D 1.495 (lit.)
bp
154-155 °C (lit.)
density
1.517 g/mL at 25 °C (lit.)
functional group
alkyl halide
iodo
SMILES string
CCCCCI
InChI
1S/C5H11I/c1-2-3-4-5-6/h2-5H2,1H3
InChI key
BLXSFCHWMBESKV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
1-Iodopentane (pentyl iodide) was used in the preparation of 1-pentyl radical by shock heating process.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
109.4 °F - closed cup
Flash Point(C)
43 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Iftikhar A Awan et al.
The journal of physical chemistry. A, 116(11), 2895-2910 (2012-02-24)
The decomposition and intramolecular H-transfer isomerization reactions of the 1-pentyl radical have been studied at temperatures of 880 to 1055 K and pressures of 80 to 680 kPa using the single pulse shock tube technique and additionally investigated with quantum
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service