Skip to Content
Merck
All Photos(2)

Key Documents

244732

Sigma-Aldrich

1-Iodo-2-methylpropane

contains copper as stabilizer, 97%

Synonym(s):

Isobutyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2I
CAS Number:
Molecular Weight:
184.02
Beilstein:
1730927
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

6 (vs air)

vapor pressure

20 mmHg ( 20 °C)

Assay

97%

form

liquid

contains

copper as stabilizer

refractive index

n20/D 1.496 (lit.)

bp

120-121 °C (lit.)

solubility

alcohol: miscible(lit.)
diethyl ether: miscible
water: insoluble(lit.)

density

1.599 g/mL at 25 °C (lit.)

functional group

alkyl halide

SMILES string

CC(C)CI

InChI

1S/C4H9I/c1-4(2)3-5/h4H,3H2,1-2H3

InChI key

BTUGGGLMQBJCBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The liquid and solid-state infrared spectra of 1-iodo-2-methylpropane were studied.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vibrational analysis of alkyl iodides: 1-iodo-2-methylpropane and 1-iodo-3-methylbutane.
Journal of Molecular Structure, 49(2), 287-292 (1978)
Xiao-Yu Yuan et al.
Bioorganic & medicinal chemistry, 27(6), 1034-1042 (2019-02-19)
Selective proteinase inhibitors have demonstrated utility in the investigation of cartilage degeneration mechanisms and may have clinical use in the management of osteoarthritis. The cysteine protease cathepsin K (CatK) is an attractive target for arthritis therapy. Here we report the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service