Skip to Content
Merck
All Photos(1)

Key Documents

8.04786

Sigma-Aldrich

1-Iodobutane

for synthesis

Synonym(s):

1-Iodobutane, Butyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3I
CAS Number:
Molecular Weight:
184.02
MDL number:
UNSPSC Code:
12352101
EC Index Number:
208-824-4
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

18.5 hPa ( 25 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

bp

130 °C/1013 hPa

mp

-103 °C

transition temp

flash point 33 °C

density

1.62 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

ICCCC

InChI

1S/C4H9I/c1-2-3-4-5/h2-4H2,1H3

InChI key

KMGBZBJJOKUPIA-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.613 - 1.617
Identity (IR): passes test

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

91.4 °F - closed cup

Flash Point(C)

33 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service