245798
2-Iodo-2-methylpropane
contains copper as stabilizer, 95%
Synonym(s):
tert-Butyl iodide
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Assay
95%
form
liquid
contains
copper as stabilizer
refractive index
n20/D 1.491 (lit.)
bp
99-100 °C (lit.)
mp
−38 °C (lit.)
density
1.544 g/mL at 25 °C (lit.)
functional group
alkyl halide
iodo
SMILES string
CC(C)(C)I
InChI
1S/C4H9I/c1-4(2,3)5/h1-3H3
InChI key
ANGGPYSFTXVERY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
2-Iodo-2-methylpropane was degraded by reductive dehalogenation with nickel-aluminum alloy in potassium hydroxide solution.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Physical chemistry chemical physics : PCCP, 23(5), 3311-3320 (2021-02-03)
We have obtained properties (or descriptors) of the transition states in the solvolysis of tert-butyl chloride, bromide and iodide. We show that all three transition states, in both protic and in aprotic solvents, are highly dipolar and are strong hydrogen
Chemistry, an Asian journal, 15(11), 1701-1708 (2020-04-16)
Alkylation of homofullerene [6,6]-C60 (CF2 )2- dianion with the set of alkyl halides, RX, was established to demonstrate an effect of RX nature on the conversion, product composition, and regioselectivity. The respective C60 (CF2 )RH, C60 (CF2 )R2 and C60
American Industrial Hygiene Association journal, 52(6), 252-257 (1991-06-01)
Two techniques were investigated for degrading a number of halogenated compounds of commercial and research importance. Reductive dehalogenation with nickel-aluminum alloy in potassium hydroxide solution was used to degrade iodomethane, chloroacetic acid, trichloroacetic acid, 2-chloroethanol, 2-bromoethanol, 2-chloroethylamine, 2-bromoethylamine, 1-bromobutane, 1-iodobutane
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service