Skip to Content
Merck
All Photos(1)

Key Documents

238252

Sigma-Aldrich

1-Iododecane

98%

Synonym(s):

Decyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9I
CAS Number:
Molecular Weight:
268.18
Beilstein:
1735228
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>5 (vs air)

vapor pressure

0.01 mmHg ( 20 °C)

Assay

98%

form

liquid

contains

copper as stabilizer

refractive index

n20/D 1.485 (lit.)

bp

132 °C/15 mmHg (lit.)

density

1.257 g/mL at 25 °C (lit.)

functional group

alkyl halide
iodo

SMILES string

CCCCCCCCCCI

InChI

1S/C10H21I/c1-2-3-4-5-6-7-8-9-10-11/h2-10H2,1H3

InChI key

SKIDNYUZJPMKFC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Electrochemical reduction of 1-iododecane at mercury cathodes in DMF containing tetra-n-butylammonium perchlorate or tetramethyl ammonium perchlorate (supporting electrolyte) has been investigated.

Application

1-Iododecane has been used in the preparation of 9-nonadecanone via palladium-catalyzed carbonylative cross-coupling reaction with 9-octyl-9-borabicyclo[3.3.l]nonane.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical reduction of 1-iododecane and 1-bromodecane at a mercury cathode in dimethylformamide.
McNamee GM, et al.
Journal of the American Chemical Society, 99(6), 1831-1835 (1977)
Niluksha Walalawela et al.
Photochemistry and photobiology, 95(5), 1160-1168 (2019-03-19)
In order to develop a new long alkane chain pterin that leaves the pterin core largely unperturbed, we synthesized and photochemically characterized decyl pterin-6-carboxyl ester (CapC) that preserves the pterin amide group. CapC contains a decyl-chain at the carboxylic acid
Palladium-catalyzed carbonylative cross-coupling reaction of iodoalkanes with 9-alkyl-9-BBN derivatives. A direct and selective synthesis of ketones.
Ishiyama T and Miyaura N.
Tetrahedron Letters, 32(47), 6923-6926 (1991)
María José Sosa et al.
Photochemistry and photobiology, 97(1), 80-90 (2020-07-07)
Mono- and bis-decylated lumazines have been synthesized and characterized. Namely, mono-decyl chain [1-decylpteridine-2,4(1,3H)-dione] 6a and bis-decyl chain [1,3-didecylpteridine-2,4(1,3H)-dione] 7a conjugates were synthesized by nucleophilic substitution (SN 2) reactions of lumazine with 1-iododecane in N,N-dimethylformamide (DMF) solvent. Decyl chain coupling occurred
Thomas Bielewicz et al.
Nanotechnology, 27(35), 355602-355602 (2016-07-28)
Two-dimensional colloidal nanosheets represent very attractive optoelectronic materials. They combine good lateral conductivity with solution-processability and geometry-tunable electronic properties. In the case of PbS nanosheets, so far synthesis has been driven by the addition of chloroalkanes as coligands. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service