Skip to Content
Merck
All Photos(1)

Key Documents

09699

Supelco

Ammonium carbamate

for decomposition

Synonym(s):

Carbamic acid ammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCO2NH4
CAS Number:
Molecular Weight:
78.07
Beilstein:
3914091
EC Number:
MDL number:
UNSPSC Code:
12000000
eCl@ss:
39033203
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

100 mmHg ( 26.7 °C)

Quality Level

Assay

≥99.5% (T)

expl. lim.

2 %

ign. residue

≤0.002% (as SO4)

anion traces

chloride (Cl-): ≤5 mg/kg
nitrate (NO3-): ≤10 mg/kg
phosphate (PO43-): ≤5 mg/kg
sulfate (SO42-): ≤10 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

storage temp.

2-8°C

SMILES string

[H]N([H])[H].NC(O)=O

InChI

1S/CH3NO2.H3N/c2-1(3)4;/h2H2,(H,3,4);1H3

InChI key

BVCZEBOGSOYJJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

this product will be discontinued, please find the alternative product here :
101134

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alfin D N Vaz et al.
Rapid communications in mass spectrometry : RCM, 24(14), 2109-2121 (2010-06-17)
A simple procedure is described to identify acyl-glucuronides by coupled liquid chromatography/mass spectrometry after derivatization to a hydroxamic acid with hydroxylamine. The reaction specificity obviates the need for isolation of the acyl-glucuronide from an extract. Glucuronides derived from carbamic acids
William Conway et al.
Environmental science & technology, 47(2), 1163-1169 (2012-11-30)
The present study reports (a) the determination of both the kinetic rate constants and equilibrium constants for the reaction of CO(2)(aq) with sterically hindered amines and (b) an attempt to elucidate a fundamental chemical understanding of the relationship between the
S Tolosa Arroyo et al.
The journal of physical chemistry. A, 113(9), 1858-1863 (2009-02-13)
A procedure is described for the theoretical study of chemical reactions in solution by means of molecular dynamics simulation, with solute-solvent interaction potentials derived from ab initio quantum calculations. We apply the procedure to the case of neutral hydrogen isocyanate
Nobuhiko Joki et al.
Clinical calcium, 22(7), 993-999 (2012-07-04)
The one of the main concept of CKD-MBD theory is to prevent and to retard progression of vascular calcification. Recently many basic researches have approached to clear the mechanism of uremic specific vascular calcification. However it remains unclear in clinical
Yannick Mamindy-Pajany et al.
Environmental science and pollution research international, 20(7), 4414-4423 (2012-12-06)
A soil column adsorption-desorption study was performed on an agricultural calcareous soil to determine the impact of sewage sludge spreading on nickel mobility. Ni adsorption experiments were followed by desorption tests involving the following liquid extractants: water, calcium (100 mg/L), oxalic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service