Merck
  • [Kidney and bone update : the 5-year history and future of CKD-MBD. Vascular calcification in CKD patients].
[Kidney and bone update : the 5-year history and future of CKD-MBD. Vascular calcification in CKD patients].
Clinical calcium (2012-07-04)
Nobuhiko Joki, Masaki Iwasaki
ABSTRACT
The one of the main concept of CKD-MBD theory is to prevent and to retard progression of vascular calcification. Recently many basic researches have approached to clear the mechanism of uremic specific vascular calcification. However it remains unclear in clinical setting what examination is best for evaluating vascular calcification. Moreover it is still not sure whether the management based on guidelines can prevent progression of vascular calcification or not in CKD patients. We review about the most accessible clinical factors for vascular calcification.
Sigma-Aldrich
Ammonium carbamate, 99%
Supelco
Ammonium carbamate, for decomposition