Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ammonium carbamate
Synonym(s):
Carbamic acid ammonium salt, Ammonium carbamate
Linear Formula:
H
2
NCO
2
NH
4
CAS Number:
1111-78-0
Molecular Weight:
78.07
EC Number:
214-185-2
Products
Product Category
salts (1)
Brand
Sigma-Aldrich (1)
Supelco (2)
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Puriss (1)
Puriss P.A. (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
292834
Ammonium carbamate,
99%
Expand
View Pricing
1.01134
Ammonium carbamate,
for analysis EMSURE
®
Expand
View Pricing
09699
Ammonium carbamate,
for decomposition
Expand
View Pricing