Merck
  • Impact of sewage sludge spreading on nickel mobility in a calcareous soil: adsorption-desorption through column experiments.
Impact of sewage sludge spreading on nickel mobility in a calcareous soil: adsorption-desorption through column experiments.
Environmental science and pollution research international (2012-12-06)
Yannick Mamindy-Pajany, Stéphanie Sayen, Emmanuel Guillon
ABSTRACT
A soil column adsorption-desorption study was performed on an agricultural calcareous soil to determine the impact of sewage sludge spreading on nickel mobility. Ni adsorption experiments were followed by desorption tests involving the following liquid extractants: water, calcium (100 mg/L), oxalic acid (525 mg/L equivalent to 100 mg carbon/L), and sludge extracts (0.5 and 2.5 g/L). Desorption tests were also conducted after sewage sludge spreading at three application rates (30, 75, and 150 t/ha). According to the breakthrough curve, Ni adsorption was irreversible and occurred mainly through interactions with calcite surface sites. Nickel desorption from the soil column was promoted in presence of significant dissolved organic carbon (DOC) concentration as observed with oxalic acid elution and sludge extract at 2.5 g/L. In sludge-amended soil columns, the maximum Ni levels occurred in first pore volumes, and they were positively correlated to the sludge application rate. The presence of DOC in leaching waters was the main factor controlling Ni desorption from the sludge-amended soil columns. This finding implies that DOC generated by sludge applied on calcareous soils might facilitate the leaching of Ni due to the formation of soluble Ni-organic complexes. Thus, sludge application can have potential environmental impacts in calcareous soils, since it promotes nickel transport by decreasing Ni retention by soil components.
Sigma-Aldrich
Ammonium carbamate, 99%
Supelco
Ammonium carbamate, for decomposition