Merck
  • A rapid and specific derivatization procedure to identify acyl-glucuronides by mass spectrometry.
A rapid and specific derivatization procedure to identify acyl-glucuronides by mass spectrometry.
Rapid communications in mass spectrometry : RCM (2010-06-17)
Alfin D N Vaz, Wei Wei Wang, Andrew J Bessire, Raman Sharma, Anne E Hagen
ABSTRACT
A simple procedure is described to identify acyl-glucuronides by coupled liquid chromatography/mass spectrometry after derivatization to a hydroxamic acid with hydroxylamine. The reaction specificity obviates the need for isolation of the acyl-glucuronide from an extract. Glucuronides derived from carbamic acids, and alkyl- and aromatic amines, are inert to the derivatization reaction conditions, making the hydroxamic acid derivative a fingerprint for acyl-glucuronides.
Sigma-Aldrich
Ammonium carbamate, 99%
Supelco
Ammonium carbamate, for decomposition