735159
Fluorine doped tin oxide coated glass slide
L × W × thickness 100 mm × 100 mm × 2.3 mm, surface resistivity ~7 Ω/sq
Synonym(s):
TEC 7, FTO glass
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
description
Haze: 5%
Quality Level
composition
SnO2/F
surface resistivity
~7 Ω/sq
L × W × thickness
100 mm × 100 mm × 2.3 mm
transmittance
80-82% (visible)
Looking for similar products? Visit Product Comparison Guide
General description
The coating thickness of Fluorine doped tin oxide on glass slide is ∼ 550 nm.The glass has a refractive index of ∼ 1.52 and the SnO2:F coating is ∼ 1.85.
Application
- Propranolol in Schizophrenia Pharmacokinetics: A meta-analysis investigates the impact of Propranolol on the pharmacokinetics of clozapine in schizophrenic patients, contributing to the understanding of beta-blockers in psychiatric medication management (Yang et al., 2024).
- Propranolol for Intraocular Delivery: Development of polymeric Propranolol nanoparticles aims at enhancing intraocular delivery, offering a new approach for controlling diseases like glaucoma and potentially improving vitreous pharmacokinetics (Chaharband et al., 2024).
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Siyeong Yang et al.
Nanomaterials (Basel, Switzerland), 9(4) (2019-04-13)
Au nanostructures (Au NSs) have been considered promising materials for applications in fuel cell catalysis, electrochemistry, and plasmonics. For the fabrication of high-performance Au NS-based electronic or electrochemical devices, Au NSs should have clean surfaces and be directly supported on
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service