Skip to Content
Merck
All Photos(2)

Documents

703176

Sigma-Aldrich

Indium tin oxide coated glass slide, square

surface resistivity 70-100 Ω/sq

Synonym(s):

ITO, ITO coated slide, square

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
In2O3 · (SnO2)x
CAS Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

description

slide

surface resistivity

70-100 Ω/sq

L × W × thickness

25 mm × 25 mm × 1.1 mm

transmittance

86% (nominal at 550nm)

refractive index

n20/D 1.517

SMILES string

O=[Sn]=O.O=[In]O[In]=O

InChI

1S/2In.5O.Sn

InChI key

LNNWKAUHKIHCKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

substrate: unpolished float glass, SiO2 passivated (200-300 Å)

Physical properties

Thickness of ITO coating is 150-300 Å

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service