735191
Fluorine doped tin oxide coated glass slide
L × W × thickness 300 mm × 300 mm × 3.2 mm, surface resistivity ~8 Ω/sq
Synonym(s):
TEC 8, FTO glass
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
description
Haze: 12%
composition
SnO2/F
surface resistivity
~8 Ω/sq
L × W × thickness
300 mm × 300 mm × 3.2 mm
transmittance
80-81.5% (visible)
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service