Skip to Content
Merck
All Photos(2)

Key Documents

576360

Sigma-Aldrich

Indium tin oxide coated boro-aluminosilicate glass slide

surface resistivity 5-15 Ω/sq, slide

Synonym(s):

ITO coated slide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
In2O3 · (SnO2)x
CAS Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

surface resistivity

5-15 Ω/sq

L × W × thickness

75 mm × 25 mm × 1.1 mm

transmittance

>85%

refractive index

n20/D 1.5290-1.5460 (lit.)

transition temp

softening point 975 °C

SMILES string

O=[Sn]=O.O=[In]O[In]=O

InChI

1S/2In.5O.Sn

InChI key

LNNWKAUHKIHCKO-UHFFFAOYSA-N

General description

Typical composition 55.0% SiO2, 7.0% B2O3, 10.4% Al2O3, 21.0% CaO, and 1.0% Na2O. Strain point of slide is 666 °C.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service