Skip to Content
Merck
All Photos(2)

Key Documents

185884

Sigma-Aldrich

N,N-Dimethylacetamide

ReagentPlus®, ≥99%

Synonym(s):

DMAc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CON(CH3)2
CAS Number:
Molecular Weight:
87.12
Beilstein:
1737614
EC Number:
MDL number:
UNSPSC Code:
12352111
eCl@ss:
39031204
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

vapor density

3 (vs air)

vapor pressure

2 mmHg ( 25 °C)
4 mmHg ( 38 °C)

product line

ReagentPlus®

Assay

≥99%

form

liquid

autoignition temp.

914 °F

expl. lim.

1.8 %, 100 °F
11.5 %, 160 °F

refractive index

n20/D 1.437 (lit.)

pH

4 (20 °C, 200 g/L)

bp

164.5-166 °C (lit.)

mp

−20 °C (lit.)

density

0.937 g/mL at 25 °C (lit.)

SMILES string

CN(C)C(C)=O

InChI

1S/C4H9NO/c1-4(6)5(2)3/h1-3H3

InChI key

FXHOOIRPVKKKFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Dimethylacetamide (DMAc) is widely used to compose the solvent mixtures for cellulose. Static and dynamic light scattering studies of the cellulose dissolved in varying concentrations of DMAc/LiCl mixtures have been reported.[1]

Application

N,N-Dimethylacetamide (DMAc) has been employed as an organic solvent for the liquid-phase exfoliation of pristine graphene from bulk graphite.[2]
It may be used to compose the solvent system along with lithium chloride (LiCl) for the dissolution of commercial rayon grade cellulose.[3] DMAc/LiCl solvent mixtures have been used for the preparation of chitin, cellulose and polyimide fibers and films.[4]
N,N-Dimethylacetamide may be used in the preparation of α-lithio N,N-dimethylacetamide.[5] DMAc along with palladium chloride forms an efficient catalytic system for the oxidation of several internal olefins to carbonyl compounds without undergoing isomerization.[6]

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wacker-Type Oxidation of Internal Olefins Using a PdCl2/N,N-Dimethylacetamide Catalyst System under Copper-Free Reaction Conditions.
Mitsudome T, et al.
Angewandte Chemie (International Edition in English), 49(7), 1238-1240 (2010)
Isolation and reactions of. alpha.-lithio N, N-dimethylacetamide.
Woodbury RP and Rathke MW
The Journal of Organic Chemistry, 42(10), 1688-1690 (1977)
B Tosh et al.
Carbohydrate research, 327(3), 345-352 (2000-08-17)
Commercial rayon grade cellulose was dissolved in the lithium chloride-N,N-dimethylacetamide (LiCl-DMAc) solvent system and esterified with acetic anhydride using p-toluenesulfonyl chloride (p-TsCl) and pyridine as catalysts. The reaction temperature was varied from 28 to 70 degrees C and the time
Transparent conductive electrodes.
YK and Chen J.
Materials for Flexible and Printed Electronics , 1(840), 6-6 (2014)
Solutions of cellulose in N, N-dimethylacetamide/lithium chloride studied by light scattering methods.
Roder T, et al.
Polymer, 42(16), 6765-6773 (2001)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service