Skip to Content
Merck
All Photos(3)

Key Documents

328634

Sigma-Aldrich

NMP

1-Methyl-2-pyrrolidinone
1 of 1 reviewers received a sample product or took part in a promotion

anhydrous, 99.5%

Synonym(s):

1-Methyl-2-pyrrolidone, N-Methyl-2-pyrrolidone, NMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO
CAS Number:
Molecular Weight:
99.13
Beilstein:
106420
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
Assay:
99.5%
bp:
202 °C (lit.)
81-82 °C/10 mmHg (lit.)
vapor pressure:
0.29 mmHg ( 20 °C)
0.99 mmHg ( 40 °C)
Pricing and availability is not currently available.

Product Name

1-Methyl-2-pyrrolidinone, anhydrous, 99.5%

grade

anhydrous

Quality Level

vapor density

3.4 (vs air)

vapor pressure

0.29 mmHg ( 20 °C)
0.99 mmHg ( 40 °C)

Assay

99.5%

form

liquid

autoignition temp.

518 °F

expl. lim.

9.5 %

impurities

<0.005% water

refractive index

n20/D 1.47 (lit.)

pH

7.7-10.0 (20 °C, 100 g/L)

bp

202 °C (lit.)
81-82 °C/10 mmHg (lit.)

mp

−24 °C (lit.)

density

1.028 g/mL at 25 °C (lit.)

SMILES string

CN1CCCC1=O

InChI

1S/C5H9NO/c1-6-4-2-3-5(6)7/h2-4H2,1H3

InChI key

SECXISVLQFMRJM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Methyl-2-pyrrolidinone may be used as a solvent in the following processes:
  • Preparation of 2-aryl and 2-alkyl benzothiazoles.[1]
  • Ullmann coupling reaction of aryl halides and phenols to form Ullmann ethers in the presence of 2,2,6,6-tetramethylheptane-3,5-dione.[2]
  • Stille cross-coupling reaction of aryl iodides with organostannanes catalyzed by Pd/C in the presence of copper iodide as cocatalyst.[3]

Other Notes

Pure-Pac® II containers require the Micromatic MacroValve coupler for dispensing solvents, Z560723.

Legal Information

Pure-Pac is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

195.8 °F - Pensky-Martens closed cup

Flash Point(C)

91 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stille couplings catalyzed by palladium-on-carbon with CuI as a cocatalyst: Synthesis of 2-(4'-acetylphenyl)thiophene
Liebeskind LS and Pe?a-Cabrera E
Organic Syntheses, 77, 135-135 (2000)
Densities, viscosities, speeds of sound, and relative permittivities for water+ cyclic amides (2-pyrrolidinone, 1-methyl-2-pyrrolidinone, and 1-vinyl-2-pyrrolidinone) at different temperatures.
George J and Sastry NV.
Journal of Chemical and Engineering Data, 49(2), 235-242 (2004)
Guohua Hu et al.
Nature communications, 8(1), 278-278 (2017-08-19)
Black phosphorus is a two-dimensional material of great interest, in part because of its high carrier mobility and thickness dependent direct bandgap. However, its instability under ambient conditions limits material deposition options for device fabrication. Here we show a black
Gerrit Homann et al.
iScience, 23(6), 101225-101225 (2020-06-21)
Frequently, poly(ethylene oxide) (PEO)-based solid polymer electrolytes (SPEs) reveal a failure with high-voltage electrodes, e.g. LiNi0.6Mn0.2Co0.2O2 in lithium metal batteries, which can be monitored as an arbitrary appearance of a "voltage noise" during charge and can be attributed to Li
Preparation of 1-methoxy-2-(4-methoxyphenoxy)benzene
Buck E and Song ZJ
Organic Syntheses, 82, 69-69 (2005)

Questions

1–4 of 4 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Does it have molecular shieve in the bottle?

    1 answer
    1. Molecular sieves are not included with this anhydrous material. However, the 18L size is packaged in the Pure-Pac II container, and the 100 mL to 2L volumes utilize Sure/Seal closures. Please see the links below to learn more about each package type:

      Sure/Seal
      https://www.sigmaaldrich.com/technical-documents/technical-article/chemistry-and-synthesis/organic-reaction-toolbox/sureseal

      Pure-Pac
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/391/402/al_techbull_al201.pdf

      Helpful?

  4. Could you please provide me with the dimensions of the bottle containing the product 328634-2L (1-Methyl-2-pyrrolidinone)?

    1 answer
    1. The packaging description for the bottle is as follows:
      • Bottle Type: Narrow Neck Glass Bottle
      • Capacity: 2500 ml (Nominal), 2750 ml (Brimful)
      • Closure Type: Crown Cap (Sure/Seal)
      • Glass Type: Amber Glass (Hydrolytic Class III)
      • Diameter: 139 mm
      • Height: 286 mm
      • Height with Cap: 293 mm
      • Thread Diameter: 32 mm
      • Thread Type: 32 mm
      • Inner Diameter of Opening: 17 mm

      Helpful?

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. boston, ma
    • Reviews 2
    • Vote 1
    4 out of 5 stars.

    nice but also

    good product, but inconsistent stock. Many times unavailable.

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service