Skip to Content
Merck
All Photos(1)

Key Documents

1601485

USP

Residual Solvent Class 2 - N,N-Dimethylacetamide

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

N,N-Dimethylacetamide solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9NO
CAS Number:
Molecular Weight:
87.12
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

N(C)(C)C(=O)C

InChI

1S/C4H9NO/c1-4(6)5(2)3/h1-3H3

InChI key

FXHOOIRPVKKKFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Residual Solvent Class 2 - N,N-Dimethylacetamide USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matija Strlic et al.
Journal of biochemical and biophysical methods, 56(1-3), 265-279 (2003-07-02)
Size exclusion of cellulose in LiCl/N,N-dimethylacetamide has been used for the past 15 years, yet much of the current research is still devoted to fundamental studies, as many issues regarding column calibration, separation mechanisms and solution behavior have not been
Mingyou Hu et al.
Organic letters, 16(7), 2030-2033 (2014-03-26)
A novel Cu-mediated trifluoromethylthiolation of diazo compounds has been developed that provides a convenient synthetic route for the efficient α-trifluoromethylthiolation of simple esters under mild reaction conditions. The reaction is typically carried out at room temperature, and water could be
Masaki Itoh et al.
Organic letters, 16(7), 2050-2053 (2014-03-22)
The direct α-methylenation of benzylpyridines was achieved using N,N-dimethylacetamide (DMA) as a one-carbon source under copper catalysis. An intermediary species was detected at an early stage, and a possible mechanism was proposed. Additionally, α-oxygenation and dimerization of benzylpyridines could also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service