Skip to Content
Merck
All Photos(3)

Key Documents

132004

Sigma-Aldrich

p-Toluenesulfonyl hydrazide

97%

Synonym(s):

p-Toluenesulfonhydrazide, p-Toluenesulfonic acid hydrazide, p-Toluenesulfonyl hydrazide, Tosylhydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO2NHNH2
CAS Number:
Molecular Weight:
186.23
Beilstein:
610130
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

103-108 °C (lit.)

functional group

hydrazine

SMILES string

Cc1ccc(cc1)S(=O)(=O)NN

InChI

1S/C7H10N2O2S/c1-6-2-4-7(5-3-6)12(10,11)9-8/h2-5,9H,8H2,1H3

InChI key

ICGLPKIVTVWCFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

p-Toluenesulfonyl hydrazide was used as a reagent for the preparation of tosylhydrazones. It was used to prepare dipyrazolo[1,5-a:4′,3′-c]pyridines and 1,2,3-selenadiazole derivatives.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - Self-react. D

Supplementary Hazards

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the American Chemical Society, 114, 966-966 (1992)
Mousa Al-Smadi et al.
Molecules (Basel, Switzerland), 13(11), 2740-2749 (2008-11-07)
The commercially available aromatic polyketones 1a-d were utilized for the synthesis of the multi-arm1,2,3-selenadiazole derivatives 3a-d. The preparation starts with the reaction between compounds 1a-d and p-toluenesulfonyl hydrazide to give the corresponding tosylhydrazones 2a-d. Subsequent reaction with selenium dioxide leads
Wolfgang Holzer et al.
Beilstein journal of organic chemistry, 8, 2223-2229 (2013-02-01)
The synthesis of dipyrazolo[1,5-a:4',3'-c]pyridines is described. Easily obtainable 5-alkynylpyrazole-4-carbaldehydes, p-toluenesulfonyl hydrazide, and an aldehyde or ketone containing an α-hydrogen atom were reacted in a silver triflate catalyzed multicomponent reaction affording new tricyclic compounds with a dipyrazolo[1,5-a:4',3'-c]pyridine core. Detailed NMR spectroscopic
Journal of the American Chemical Society, 115, 2473-2473 (1993)
D H Blakey et al.
Toxicology and industrial health, 8(6), 369-376 (1992-11-01)
Toluenesulfonhydrazide (TSH) is a high volume production chemical for which there is relatively little toxicological data. In this study, the mutagenic activity of TSH was determined in the Salmonella/mammalian microsome assay and the in vitro chromosomal aberration assay using Chinese

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service