Skip to Content
Merck
All Photos(2)

Documents

137529

Sigma-Aldrich

N,N-Diethylacetamide

97%

Synonym(s):

Acetic acid diethylamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CON(C2H5)2
CAS Number:
Molecular Weight:
115.17
Beilstein:
1209428
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.440 (lit.)

bp

182-186 °C (lit.)

density

0.925 g/mL at 25 °C (lit.)

SMILES string

CCN(CC)C(C)=O

InChI

1S/C6H13NO/c1-4-7(5-2)6(3)8/h4-5H2,1-3H3

InChI key

AJFDBNQQDYLMJN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Diethylacetamide is commonly used as drug solvent.

Application

N,N-Diethylacetamide was used to investigate the spectroscopic properties of N,N-diethyl-2-[(4-substituted)phenylsulfinyl] acetamides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Budden et al.
Arzneimittel-Forschung, 28(9), 1571-1579 (1978-01-01)
The drug solvents diethyleneglycol monoethylether (Transcutol), N,N-diethylacetamide, and dimethylsulfoxide were examined for their pharmacodynamic properties in the following tests: i.p. toxicity, "sign pattern", inclined screen test, balance rod test, and potentiation of hexobarbitone sleeping time in mice, spasmolytic activity in
Spectroscopic and theoretical studies of some N,N-diethyl-2-[(4-substituted) phenylsulfinyl] acetamides.
Olivato PR, et al.
Journal of Molecular Structure, 827(1), 25-34 (2007)
M Silvia et al.
Biochemical pharmacology, 48(4), 717-726 (1994-08-17)
This study was undertaken to investigate: (1) the effect of N,N-diethylacetamide (DEAC) and N,N-dimethylacetamide (DMAC) administration to rats on drug-metabolizing enzymes in the liver; (2) the in vitro dealkylation of DEAC and DMAC by hepatic microsomes from rats treated with
Action of N,N-diethylacetamide on hepatic microsomal drug-metabolizing enzymes.
F E Beyhl et al.
Food and cosmetics toxicology, 19(5), 627-629 (1981-10-01)
Vijay Gupta et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(12), 2658-2665 (2019-12-04)
A new triazole-functionalized tetracarboxylic acid ligand (H4 L) has been synthesized and utilized for the fabrication of a 3D ZnII organic framework with a Zn4 (-COO)6 cluster as the secondary building unit. The framework exhibits very good thermal stability and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service