Skip to Content
Merck
All Photos(2)

Key Documents

C4881

Sigma-Aldrich

Chlortetracycline hydrochloride

≥91.0% dry basis (HPLC)

Synonym(s):

7-Chlorotetracycline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H23ClN2O8 · HCl
CAS Number:
Molecular Weight:
515.34
Beilstein:
3858364
EC Number:
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85

Quality Level

Assay

≥91.0% dry basis (HPLC)

form

powder

pKa (25 °C)

3.3
7.4
9.3

mp

210-215 °C (dec.) (lit.)

solubility

1 M NaOH: 50 mg/mL

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

protein synthesis | interferes

storage temp.

−20°C

SMILES string

Cl.CN(C)[C@H]1[C@@H]2CC3C(=C(O)[C@]2(O)C(=O)C(C(N)=O)=C1O)C(=O)c4c(O)ccc(Cl)c4[C@@]3(C)O

InChI

1S/C22H23ClN2O8.ClH/c1-21(32)7-6-8-15(25(2)3)17(28)13(20(24)31)19(30)22(8,33)18(29)11(7)16(27)12-10(26)5-4-9(23)14(12)21;/h4-5,7-8,15,26,28-29,32-33H,6H2,1-3H3,(H2,24,31);1H/t7?,8-,15-,21-,22-;/m0./s1

InChI key

CBHYYLPALVVVEY-LYNLVHCPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: tetracycline

Application

Chlortetracycline was used in fluorescence assays to detect the Ca+2 influx required for the completion of acrosome reaction in human spermatozoa.

Biochem/physiol Actions

Chlortetracycline is an antibiotic produced by some strains of Streptomyces aureofaciens. It inhibits protein synthesis (elongation) by preventing binding of aminoacyl-tRNA to the 30S subunit. It is effective against Gram-positive and to a lesser degree Gram-negative bacteria than tetracycline.
Mode of Resistance: Loss of cell wall permeability.

Other Notes

If stored frozen, chlortetracycline hydrochloride is expected to remain stable at least four years.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W K Amoa-Awua et al.
Journal of applied microbiology, 83(3), 288-296 (1997-11-14)
Agbelima, a fermented cassava meal widely consumed in Ghana, Togo and Benin, is produced by fermenting grated cassava with one of several types of traditional cassava dough inoculum. During fermentation a smooth textured sour dough is produced, the toxicity of
A H Amin et al.
Human reproduction (Oxford, England), 11(4), 741-745 (1996-04-01)
This study was designed to compare three different fluorescent probes to assay the acrosome reaction in human spermatozoa: chlortetracycline (CTC), mannosylated bovine serum albumin (BSA) labelled with fluorescein (MAF), and quinacrine (QN). Normal human sperm ejaculates were washed and allowed
Y H Choi et al.
Biology of reproduction, 59(6), 1328-1333 (1998-11-26)
Cyclodextrin, which stimulates cholesterol efflux from cells, was examined for its ability to induce capacitation of mouse spermatozoa. A chemically defined, protein-free medium was used for in vitro fertilization of cumulus-free mouse eggs. Fertilization did not occur in modified Krebs-Ringer
A E Oliver et al.
Biophysical journal, 78(4), 2116-2126 (2000-03-29)
The effect of temperature on the binding equilibria of calcium-sensing dyes has been extensively studied, but there are also important temperature-related changes in the photophysics of the dyes that have been largely ignored. We conducted a systematic study of thermal
Jasmer Dalal et al.
Scientific reports, 9(1), 6712-6712 (2019-05-02)
The objective of this study was to determine the mechanism by which RU 486 (mifepristone) protects sperm to undergo premature capacitation during cryopreservation. For this, semen ejaculate (n = 20) was divided into four equal fractions and diluted using egg yolk-based extender

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service