Skip to Content
Merck

Skip To

O4636

Oxytetracycline dihydrate

Assay (anhydrous basis) 95.0%-102.0%

Synonym(s):

5-Hydroxytetracycline dihydrate, Oxytetracycline, Oxytetracycline Anhydrous

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C22H24N2O9 · 2H2O
CAS Number:
Molecular Weight:
496.46
NACRES:
NA.85
PubChem Substance ID:
UNSPSC Code:
51284038
EC Number:
201-212-8
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria

mode of action

protein synthesis | interferes

SMILES string

O.[H][C@@]12[C@@H](O)[C@@]3([H])C(C(=O)c4c(O)cccc4[C@@]3(C)O)=C(O)[C@]1(O)C(=O)C(C(N)=O)=C(O)[C@H]2N(C)C

InChI

1S/C22H24N2O9.H2O/c1-21(32)7-5-4-6-8(25)9(7)15(26)10-12(21)17(28)13-14(24(2)3)16(27)11(20(23)31)19(30)22(13,33)18(10)29;/h4-6,12-14,17,25,27-29,32-33H,1-3H3,(H2,23,31);1H2/t12-,13-,14+,17+,21-,22+;/m1./s1

InChI key

IBZHEBHGZFICKS-IFLJXUKPSA-N

General description

Chemical structure: tetracycline

Application

Oxytetracycline is a tetracycline analog isolated from the actinomycete Streptomyces rimosus. It is used in studies on the oxytetracycline-resistance gene (otrA) and is used for fluorescence measurements of hepatocytes[1].

Biochem/physiol Actions

Antibiotic produced by Streptomyces rimosus.
Mode of Action: Inhibits protein synthesis (elongation) by preventing binding of aminoacyl-tRNA to the 30S subunit.
Antimicrobial spectrum: Gram-negative and Gram-positive bacteria.
Mode of Resistance: Active efflux, ribosome protection, tetracycline inactivation.
Oxytetracycline inhibits translation, which results in cell death. It binds to the 30S ribosomal subunit and prevents the amino-acyl tRNA from binding to the A site of the ribosome, inhibiting protein synthesis (elongation). The binding is reversible in nature. Oxytetracycline is lipophilic so it can easily pass through a cell membrane and passively diffuses through channels in the bacterial membrane.

Analysis Note

solubility very slightly solube in water, it dissolves in dilute acid and alkaline solutions

Other Notes

10g,50g
Keep container tightly closed in a dry and well-ventilated place.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T3258O5875T8032
antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

mode of action

protein synthesis | interferes

mode of action

protein synthesis | interferes

mode of action

protein synthesis | interferes

mode of action

protein synthesis | interferes


pictograms

Health hazardEnvironment

signalword

Warning

Hazard Classifications

Aquatic Chronic 2 - Repr. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters