Skip to Content
Merck
All Photos(1)

Documents

33700

Supelco

Danofloxacin

VETRANAL®, analytical standard

Synonym(s):

Advocin®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20FN3O3
CAS Number:
Molecular Weight:
357.38
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

CN1C[C@@H]2C[C@H]1CN2c3cc4N(C=C(C(O)=O)C(=O)c4cc3F)C5CC5

InChI

1S/C19H20FN3O3/c1-21-7-12-4-11(21)8-22(12)17-6-16-13(5-15(17)20)18(24)14(19(25)26)9-23(16)10-2-3-10/h5-6,9-12H,2-4,7-8H2,1H3,(H,25,26)/t11-,12-/m0/s1

InChI key

QMLVECGLEOSESV-RYUDHWBXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

Advocin is a registered trademark of Pfizer, Inc.
VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mariana Ballent et al.
Veterinary journal (London, England : 1997), 192(3), 422-427 (2011-11-15)
The impact of an efflux pump-related interaction between ivermectin and danofloxacin on their intestinal transport (ex vivo) and disposition kinetics (in vivo) was assessed. Eighteen male Corriedale sheep were randomly assigned to one of three groups. Animals in Group A
Ana García-Galán et al.
Pathogens (Basel, Switzerland), 9(7) (2020-07-11)
Mycoplasma bovis is an important bovine pathogen causing pneumonia, mastitis, and arthritis and is responsible for major economic losses worldwide. In the absence of an efficient vaccine, control of M. bovis infections mainly relies on antimicrobial treatments, but resistance is
Andrea Speltini et al.
Analytical and bioanalytical chemistry, 404(5), 1565-1569 (2012-07-21)
Here we describe the extraction from soil of the major photo-transformation products (PTPs) of enrofloxacin (ENR) and danofloxacin (DAN), two fluoroquinolones (FQs) widely used in veterinary medicine and of growing environmental concern, because their PTPs have been shown to retain
Guoying Chen et al.
Journal of agricultural and food chemistry, 59(4), 1058-1062 (2011-01-14)
Danofloxacin (DANO) residue in bovine muscle was screened at 200 ng/g by terbium-sensitized luminescence (TSL) directly measured on 10 × 6 mm C18 sorbent strips. The analyte was first adsorbed on sorbent surface by immersion in defatted homogenates. After reagent
Xuan-Anh Ton et al.
Biosensors & bioelectronics, 36(1), 22-28 (2012-05-01)
A rapid, robust, sensitive and economic sensing method, based on a molecularly imprinted polymer (MIP) synthetic antibody mimic, and fluorescence polarization analysis, for the direct detection of UV-excited fluorescent analytes in food and environmental samples was developed. Fluoroquinolone (FQ) antibiotics

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service