Skip to Content
Merck
All Photos(1)

Documents

792632

Sigma-Aldrich

Isoflurane

Synonym(s):

Isoflurane, 1-Chloro-1-(difluoromethoxy)-2,2,2-trifluoroethane, 1-Chloro-2,2,2-trifluoroethyl difluoromethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H2ClF5O
CAS Number:
Molecular Weight:
184.49
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

238 mmHg ( 20 °C)

form

liquid

refractive index

n20/D 1.302

bp

48.5 °C

density

1.510 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

FC(F)OC(Cl)C(F)(F)F

InChI

1S/C3H2ClF5O/c4-1(3(7,8)9)10-2(5)6/h1-2H

InChI key

PIWKPBJCKXDKJR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isofluraneis a fluorinated methyl ether and isomer of enflurane. The Federal DrugAdministration (FDA) has approved it for use as a volatile anesthetic to induceand maintain general anesthesia.

Application

Isoflurane is a volatile anesthetic agent mainly used to study its interaction with the biological system.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT SE 3

Target Organs

Central nervous system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isoflurane-induced neuroapoptosis in the neonatal rhesus macaque brain.
Brambrink AM, et al.
Anesthesiology, 112(4), 834-841 (2010)
Satya Prathyusha Bhamidimarri et al.
Structure (London, England : 1993), 27(2), 268-280 (2018-12-18)
Research efforts to discover potential new antibiotics for Gram-negative bacteria suffer from high attrition rates due to the synergistic action of efflux systems and the limited permeability of the outer membrane (OM). One strategy to overcome the OM permeability barrier
Wenjuan Sun et al.
Frontiers in pharmacology, 12, 708575-708575 (2021-10-01)
Maslinic acid (MA), also named crategolic acid, is a pentacyclic triterpene extracted from fruits and vegetables. Although various beneficial pharmacological effects of MA have been revealed, its effect on renal fibrosis remains unclear. This study was designed to clarify whether
Hao Zhang et al.
Molecular medicine reports, 22(2), 915-925 (2020-07-07)
Dilated cardiomyopathy (DCM) is a primary cardiomyopathy with high mortality. The aim of the present study was to identify the related genes in DCM. The four expression profiles (GSE17800, GSE21610, GSE42955 and GSE79962) downloaded from the Gene Expression Omnibus (GEO)
Sherrefa R Burchell et al.
Journal of investigative medicine : the official publication of the American Federation for Clinical Research, 61(7), 1078-1083 (2013-07-26)
Isoflurane is a volatile anesthetic that is widely used clinically as an inhalational anesthetic. In recent years, several studies have indicated that isoflurane has neuroprotective properties. This has led to the beneficial effects of isoflurane being analyzed in both cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service