Skip to Content
Merck

1303501

USP

Halothane

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-Bromo-2-chloro-1,1,1-trifluoroethane, Halothane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
BrCH(Cl)CF3
CAS Number:
Molecular Weight:
197.38
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1736947
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

vapor pressure

4.5 psi ( 20 °C)

API family

halothane

manufacturer/tradename

USP

refractive index

n20/D 1.369 (lit.)

bp

50.2 °C (lit.)

density

1.872 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

FC(F)(F)C(Cl)Br

InChI

1S/C2HBrClF3/c3-1(4)2(5,6)7/h1H

InChI key

BCQZXOMGPXTTIC-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Halothane USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Biochem/physiol Actions

2-Bromo-2-chloro-1,1,1-trifluoroethane is an inhalation anesthetic. In an effect believed to be independent of anesthesia, halothane inhibits the synthesis of 5-hydroxytryptamine in brain tissue, probably at the tryptophan hydroxylase step.
Inhalation anesthetic.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


signalword

Danger

Hazard Classifications

Eye Dam. 1 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library