Skip to Content
Merck
All Photos(1)

Key Documents

455008

Sigma-Aldrich

Poly(ethylene glycol) diacrylate

average Mn 700, acrylate, 100 ppm MEHQ as inhibitor, 300 ppm BHT as inhibitor

Synonym(s):

Polyethylene glycol, PEG diacrylate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Product Name

Poly(ethylene glycol) diacrylate, average Mn 700

mol wt

average Mn 700

contains

100 ppm MEHQ as inhibitor (typically)
300 ppm BHT as inhibitor (typically)

reaction suitability

reagent type: cross-linking reagent
reaction type: Polymerization Reactions

refractive index

n20/D 1.47

mp

12-17 °C

density

1.12 g/mL at 25 °C

Ω-end

acrylate

α-end

acrylate

polymer architecture

shape: linear
functionality: homobifunctional

storage temp.

2-8°C

SMILES string

OCCO.OC(=O)C=C

InChI

1S/C8H10O4/c1-3-7(9)11-5-6-12-8(10)4-2/h3-4H,1-2,5-6H2

InChI key

KUDUQBURMYMBIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene glycol) diacrylate (PEGDA) is a derivative of polyethylene glycol that can be used for a variety of drug delivery and tissue engineering based applications. It is used as a prepolymer solution that can be used in the formation of a cross-linked polymeric system.

Application

PEGDA can be used in the formation of sulphonated ion exchange membranes for fuel cell applications. It may also be used in the development of novel injectable biodegradable polymers for a variety of biomedical applications.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

446.0 °F - closed cup

Flash Point(C)

230 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sodium chloride sorption in sulfonated polymers for membrane applications
Geise GM, et al.
Journal of Membrane Science , 423(9), 195-208 (2012)
Photopolymerizable sulfonated poly (ethylene glycol) proton exchange membranes for microfluidic and fuel cell applications
Nearingburg B and Elias AL
Journal of Membrane Science , 389, 148-154 (2012)
Ho-Joon Lee et al.
Biotechnology and bioengineering, 117(6), 1864-1876 (2020-03-13)
Although primary human hepatocytes (PHHs) are the gold standard in drug efficacy and metabolism studies, long-term survival of PHHs and maintenance of their hepatic function are still challenging. In this study, we focused on the effect of the initial microenvironment
Injectable biodegradable polymer composites based on poly (propylene fumarate) crosslinked with poly (ethylene glycol)-dimethacrylate
He S, et al.
Biomaterials, 21(23), 2389-2394 (2000)
Yi-Sin Chen et al.
Lab on a chip, 19(10), 1764-1771 (2019-04-04)
An integrated microfluidic system combining 1) an optically-induced-dielectrophoresis (ODEP) module for manipulation of drug-containing particles and 2) an ultraviolet (UV) "direct writing" module capable of patterning hydrogels was established herein for automatic formulation of customized digital drug cocktails. Using the

Articles

Scaffold patterning with poly(ethylene glycol)-based hydrogels for cell presence in 2D and 3D environments on photoactive substrates.

Scaffold patterning with poly(ethylene glycol)-based hydrogels for cell presence in 2D and 3D environments on photoactive substrates.

Scaffold patterning with poly(ethylene glycol)-based hydrogels for cell presence in 2D and 3D environments on photoactive substrates.

Scaffold patterning with poly(ethylene glycol)-based hydrogels for cell presence in 2D and 3D environments on photoactive substrates.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service