Skip to Content
Merck
All Photos(1)

Key Documents

T8802

Sigma-Aldrich

Trypsin from bovine pancreas

TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

biological source

bovine pancreas

Quality Level

sterility

aseptically filled

form

essentially salt-free, lyophilized powder

specific activity

≥10,000 BAEE units/mg protein

mol wt

23.8 kDa

composition

protein, ≥95%

solubility

hydrochloric acid: soluble 1 mM, clear

foreign activity

Chymotrypsin ≤0.1 BTEE units/mg protein

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

The trypsin molecule has two domains: one is related to the enzyme active site and the tryptophan residues; the other is related to the 8-anilinonaphthalene-1-sulfonate binding.

Application

For trypsin digestion of peptides, use a ratio of about 1:100 to 1:20 for trypsin:peptide. The typical use for this product is in removing adherent cells from a culture surface. The concentration of trypsin necessary to dislodge cells from their substrate is dependent primarily on the cell type and the age of the culture. Trypsins have also been used for the re-suspension of cells during cell culture, in proteomics research for digestion of proteins and in various in-gel digestions. Additional applications include assessing crystallization by membrane-based techniques and in a study to determine that protein folding rates and yields can be limited by the presence of kinetic traps.
Trypsin can be used to release adherent cells from tissue culture plates for passaging. Trypsin has been used in a study to assess the effects of macromolecular crowding on the structural stability of human α-lactalbumin. [1] Trypsin has also been used in a study to investigate BN-PAGE analysis of Trichoderma harzianum secretome. [2]

Biochem/physiol Actions

Trypsin cleaves peptides on the C-terminal side of lysine and arginine residues. The rate of hydrolysis of this reaction is slowed if an acidic residue is on either side of the cleavage site and hydrolysis is stopped if a proline residue is on the carboxyl side of the cleavage site. The optimal pH for trypsin activity is 7-9. Trypsin can also act to cleave ester and amide linkages of synthetic derivatives of amino acids. EDTA is added to trypsin solutions as a chelating agent that neutralizes calcium and magnesium ions that obscure the peptide bonds on which trypsin acts. Removing these ions increases the enzymatic activity.

Serine protease inhibitors, including DFP, TLCK, APMSF, AEBSEF, and aprotinin, amongst others, will inhibit Trypsin.

Unit Definition

One BAEE unit will produce a ΔA253 of 0.001 per min at pH 7.6 at 25 °C using BAEE as substrate. Reaction volume = 3.2 ml (1 cm light path).
One BAEE unit will produce a A253 of 0.001 per minute at pH 7.6 at 25°C using BAEE as a substrate.

Preparation Note

TPCK treated

Analysis Note

Protein determined by E1%/280

Other Notes

View more information on trypsin at www.sigma-aldrich.com/enzymeexplorer

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adelson Joel da Silva et al.
Proteomics, 12(17), 2729-2738 (2012-06-30)
Plant cell wall-degrading enzymes produced by microorganisms possess important biotechnological applications, including biofuel production. Some anaerobic bacteria are able to produce multienzymatic complexes called cellulosomes while filamentous fungi normally secrete individual hydrolytic enzymes that act synergistically for polysaccharide degradation. Here
Piliang Hao et al.
Journal of proteome research, 14(2), 1308-1314 (2014-12-17)
Nonenzymatic deamidation occurs readily under the condition of trypsin digestion, resulting in the identification of many artificial deamidation sites. To evaluate the effect of trypsin digestion buffers on artificial deamidation, we compared the three commonly used buffers Tris-HCl (pH 8)
De-Lin Zhang et al.
Acta biochimica et biophysica Sinica, 44(8), 703-711 (2012-06-28)
The folding of protein, an important process for protein to fulfill normal functions, takes place in crowded physiological environments. α-Lactalbumin, as a model system for protein-folding studies, has been used extensively because it can form stable molten globule states under
John C Tran et al.
Analytical chemistry, 80(5), 1568-1573 (2008-01-31)
Although well-established as a technique for protein purification, the application of continuous elution tube gel electrophoresis to proteome fractionation remains problematic. Difficulties associated with sample collection, particularly at the high mass range or at low sample loadings, continue to plague
A E M Jørgensen et al.
Osteoarthritis and cartilage, 30(6), 886-895 (2022-04-01)
Cartilage collagen has very limited repair potential, though some turnover and incorporation has not been fully excluded. We aim to determine the regional turnover of human osteoarthritis cartilage. Patients scheduled for knee joint replacement surgery due to osteoarthritis were recruited

Protocols

Continuous spectrophotometric rate determination method using BAEE substrate measures trypsin activity, essential for enzyme characterization.

Continuous spectrophotometric rate determination method using BAEE substrate measures trypsin activity, essential for enzyme characterization.

Continuous spectrophotometric rate determination method using BAEE substrate measures trypsin activity, essential for enzyme characterization.

Continuous spectrophotometric rate determination method using BAEE substrate measures trypsin activity, essential for enzyme characterization.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service