T6763
Trypsin Acetylated from bovine pancreas
Type V-S, ≥8,500 BAEE units/mg protein (biuret)
Synonym(s):
Acetyltrypsin
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Recommended Products
biological source
bovine pancreas
Quality Level
type
Type V-S
form
powder
specific activity
≥8,500 BAEE units/mg protein (biuret)
UniProt accession no.
storage temp.
−20°C
Gene Information
cow ... TRYP8(282603)
Related Categories
General description
The trypsin molecule has two domains: one is related to the enzyme active site and the tryptophan residues; the other is related to the 8-anilinonaphthalene-1-sulfonate binding.
Application
Trypsin has been used in a study to assess the ontogeny of the endocrine pancreas in the fetal/newborn baboon. Trypsin digestion and hydrogen/deuterium exchange support the transition between inward- and outward-facing conformations during the catalytic cycle of the bacterial multidrug ATP-binding cassette transporter.
Biochem/physiol Actions
Lys-acetylated trypsin is less susceptible to autolysis than the native form.
Unit Definition
One BAEE unit will produce a ΔA253 of 0.001 per min at pH 7.6 at 25° C using BAEE as substrate. Reaction volume = 3.2 mL (1 cm light path).
Preparation Note
Prepared by the method of Labouesse, J. and Gervais, M., Eur. J. Biochem., 2, 215 (1967).
Other Notes
View more information on trypsin at www.sigma-aldrich.com/enzymeexplorer.
inhibitor
Product No.
Description
Pricing
substrate
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Shin-Ichi Nakakita et al.
Methods in molecular biology (Clifton, N.J.), 2132, 567-583 (2020-04-20)
We describe a method to detect influenza virus using an evanescent-field-activated fluorescence scanner type glycan array and ELISA system. Neoglycoprotein was prepared by combination of organic chemistry and biomaterial preparation. These ligands were spotted on a glass plate or plastic
Amy R Quinn et al.
The Journal of endocrinology, 214(3), 289-299 (2012-06-23)
Erratic regulation of glucose metabolism including hyperglycemia is a common condition in premature infants and is associated with increased morbidity and mortality. The objective of this study was to examine histological and ultrastructural differences in the endocrine pancreas in fetal
Shahid Mehmood et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(27), 10832-10836 (2012-06-20)
The study of membrane proteins remains a challenging task, and approaches to unravel their dynamics are scarce. Here, we applied hydrogen/deuterium exchange (HDX) coupled to mass spectrometry to probe the motions of a bacterial multidrug ATP-binding cassette (ABC) transporter, BmrA
R F Regan et al.
Journal of neurochemistry, 70(1), 77-85 (1998-01-09)
The effect of magnesium on the oxidative neuronal injury induced by hemoglobin was assessed in murine cortical cell cultures. Exposure to 5 microM hemoglobin in physiologic (1 mM) magnesium for 26 h resulted in the death of about one-half the
Caroline Chupin et al.
Vaccines, 9(10) (2021-10-27)
The development of a live-attenuated vaccine (LAV) for the prevention of human metapneumovirus (HMPV) infection is often hampered by the lack of highly efficient and scalable cell-based production systems that support eventual global vaccine production. Avian cell lines cultivated in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service