Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
description
RTECS - YN5075000
Merck USA index - 14, 9795
Quality Segment
form
solid
specific activity
≥2,500 USP units/mg dry wt
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze, desiccated (hygroscopic)
solubility
HCl: 1 mM, neutral buffer: soluble, water: soluble
foreign activity
Chymotrypsin ≤50 NF units/mg dry wt.
shipped in
ambient
storage temp.
−20°C
General description
Physical form
Preparation Note
Other Notes
Kostka, V., and Carpenter, F.H. 1964. J. Biol. Chem.239, 1799.
Hummel, B.C.W. 1955. Can. J. Biochem. Physiol.37, 1393.
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| form solid | form tablet | form lyophilized powder | form salt-free, lyophilized powder |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| shipped in ambient | shipped in - | shipped in ambient | shipped in - |
| Quality Level 100 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| storage condition OK to freeze, desiccated (hygroscopic) | storage condition - | storage condition - | storage condition - |
| specific activity ≥2,500 USP units/mg dry wt | specific activity 90-110% (compared to standard) | specific activity 1,000-2,000 BAEE units/mg solid | specific activity - |
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.





