Skip to Content
Merck
All Photos(1)

Key Documents

B7632

Sigma-Aldrich

N-Benzoyl-Phe-Val-Arg-p-nitroanilide hydrochloride

chromogenic, protease substrate, ≥98% (TLC), powder

Synonym(s):

N-Benzoyl-L-phenylalanyl-L-valyl-L-arginine-4-nitroanilide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H40N8O6 · HCl
CAS Number:
Molecular Weight:
681.18
Beilstein:
3027332
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

N-Benzoyl-Phe-Val-Arg-p-nitroanilide hydrochloride, protease substrate

Assay

≥98% (TLC)

form

powder

solubility

methanol: 20 mg/mL, clear, colorless to light yellow

storage temp.

−20°C

SMILES string

Cl.CC(C)[C@H](NC(=O)[C@H](Cc1ccccc1)NC(=O)c2ccccc2)C(=O)N[C@@H](CCCNC(N)=N)C(=O)Nc3ccc(cc3)[N+]([O-])=O

InChI

1S/C33H40N8O6.ClH/c1-21(2)28(40-31(44)27(20-22-10-5-3-6-11-22)39-29(42)23-12-7-4-8-13-23)32(45)38-26(14-9-19-36-33(34)35)30(43)37-24-15-17-25(18-16-24)41(46)47;/h3-8,10-13,15-18,21,26-28H,9,14,19-20H2,1-2H3,(H,37,43)(H,38,45)(H,39,42)(H,40,44)(H4,34,35,36);1H/t26-,27-,28-;/m0./s1

InChI key

PYVSMZDQQUZGPF-JAQKLANPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Benzoyl-Phe-Val-Arg-p-nitroanilide is a chromogenic protease substrate.

Application

  • N

  • -Benzoyl-Phe-Val-Arg-p-nitroanilide hydrochloride has been used: as a substrate: for trypsin-like enzyme in the soluble and particulate fractions of the hyphae
  • for the thrombin, recombinant and native batroxobin from snake venom
  • for fibrinolytic enzyme aprE2 in amidolytic activity assay

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Substrates

A substrate for trypsin, thrombin and reptilase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P C Ring et al.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology, 30(8), 1085-1096 (2000-08-10)
Fel d 1, an important allergen from domestic cats, is a significant cause of asthma. In addition to directly promoting IgE synthesis, other biological activities of allergens may contribute to either allergic sensitization or the magnitude of allergic effector responses.
Substrates for determination of trypsin, thrombin and thrombin-like enzymes.
L Svendsen et al.
Folia haematologica (Leipzig, Germany : 1928), 98(4), 446-454 (1972-01-01)
Seon-Ju Jeong et al.
Journal of microbiology and biotechnology, 24(7), 969-978 (2014-04-20)
The aprE2 gene with its prosequence from Bacillus subtilis CH3-5 was overexpressed in Escherichia coli BL21(DE3) by using plasmid pET26b(+). After IPTG induction, active and mature AprE2 was produced when cells were grown at 20°C, whereas inactive and insoluble enzyme
Maxsuell Lucas Mendes Marques et al.
Marine drugs, 17(1) (2018-12-24)
In this study, sulfated polysaccharide-rich extracts were isolated from 22 tropical seaweeds (4 red, 11 brown, and 7 green) found in northeastern Brazil, and evaluated for the role of anticoagulant agents. Fifteen of the extracts showed anticoagulant activity, including all
I J Mackie et al.
Thrombosis research, 28(4), 499-507 (1982-11-15)
The Factor VIII content of Factor IX concentrates was investigated by agarose gel electrophoresis which removed the interfering effects of stabilisers and proteolytic enzyme inhibitors. Factor VIII coagulant activity (FVIII C) as measured by clotting and amidolytic methods correlated well

Protocols

Thrombin is an endolytic serine protease that selectively cleaves the Arg–Gly bonds of fibrinogen to form fibrin and release fibrinopeptides A and B.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service