Skip to Content
Merck
All Photos(1)

Key Documents

B4153

Sigma-Aldrich

Boc-Gln-Ala-Arg-7-amido-4-methylcoumarin hydrochloride

~95%

Synonym(s):

Boc-Gln-Ala-Arg-AMC hydrochloride, Boc-Gln-Ala-Arg-Mca hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H42N8O8 · HCl
CAS Number:
Molecular Weight:
667.15
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Assay

~95%

form

powder

solubility

methanol: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

CC(NC(=O)C(CCC(N)=O)NC(=O)OC(C)(C)C)C(=O)NC(CCCNC(N)=N)C(=O)Nc1ccc2C(C)=CC(=O)Oc2c1

InChI

1S/C29H42N8O8/c1-15-13-23(39)44-21-14-17(8-9-18(15)21)35-26(42)19(7-6-12-33-27(31)32)36-24(40)16(2)34-25(41)20(10-11-22(30)38)37-28(43)45-29(3,4)5/h8-9,13-14,16,19-20H,6-7,10-12H2,1-5H3,(H2,30,38)(H,34,41)(H,35,42)(H,36,40)(H,37,43)(H4,31,32,33)

InChI key

LQSLBVXESNRILG-UHFFFAOYSA-N

General description

Boc-Gln-Ala-Arg-7-amido-4-methylcoumarin hydrochloride is a fluorogenic and cell-permeable trypsin substrate.

Application

Boc-Gln-Ala-Arg-7-amido-4-methylcoumarin hydrochloride has been used as a fluorogenic substrate to measure the enzymatic activity of trypsin.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Substrates

Substrate for trypsin

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diel food intake and digestive enzyme production patterns in Solea senegalensis larvae
Navarro-Guillen C, et al.
Aquaculture (Amsterdam, Netherlands), 435, 33-42 (2015)
Evaluation of fluorogenic substrates in the assessment of digestive enzymes in a decapod crustacean Maja brachydactyla larvae
Rotllant G, et al.
Aquaculture (Amsterdam, Netherlands), 282(1-4), 90-96 (2008)
Morphological and physiological changes of Octopus bimaculoides: from embryo to juvenile
Ibarra-Garcia LE, et al.
Aquaculture (Amsterdam, Netherlands), 497, 364-372 (2018)
Sudarshan R Malla et al.
Cellular and molecular life sciences : CMLS, 77(9), 1811-1825 (2019-08-01)
Premature intrapancreatic trypsinogen activation is widely regarded as an initiating event for acute pancreatitis. Previous studies have alternatively implicated secretory vesicles, endosomes, lysosomes, or autophagosomes/autophagolysosomes as the primary site of trypsinogen activation, from which a cell-damaging proteolytic cascade originates. To
Dodheri Syed Samiulla et al.
Organic and medicinal chemistry letters, 2(1), 27-27 (2012-07-17)
Caspase-3 inhibition has been demonstrated to be therapeutically effective in moderating excessive programmed cell death. Interest in caspase-3 as a therapeutic target has led many to pursue the development of inhibitors. To date, only a few series of non-peptide inhibitors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service