Skip to Content
Merck
All Photos(1)

Documents

M7200

Sigma-Aldrich

Mesitylene

98%

Synonym(s):

1,3,5-Trimethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3(CH3)3
CAS Number:
Molecular Weight:
120.19
Beilstein:
906806
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor density

4.1 (vs air)

Quality Level

vapor pressure

14 mmHg ( 55 °C)
2.49 mmHg ( 25 °C)

Assay

98%

form

liquid

autoignition temp.

1022 °F

expl. lim.

0.88-6.1 %, 100 °F

refractive index

n20/D 1.499 (lit.)

bp

163-166 °C (lit.)

mp

−45 °C (lit.)

density

0.864 g/mL at 25 °C (lit.)

SMILES string

Cc1cc(C)cc(C)c1

InChI

1S/C9H12/c1-7-4-8(2)6-9(3)5-7/h4-6H,1-3H3

InChI key

AUHZEENZYGFFBQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Mesitylene, also known as 1,3,5-trimethylbenzene, is a benzene derivative. Its carbon-13 spin-lattice relaxation times, dipolar relaxation rates and nuclear Overhauser enhancements have been measured to analyze the internal methyl rotation and intermolecular interactions. The anodic oxidation of mesitylene at a platinum electrode in the presence of different electroytes has been investigated. The ring deformation in mesitylene has been studied in the gas-phase by electron diffraction.

Application

Mesitylene may be used to:
  • Grow fullerene(C60)/1,3,5-trimethylbenzene (mesitylene) nanowires.
  • Prepare 2′,4′,6′-trimethyl acetophenone with high slectivity by reacting with acetyl chloride in the presence of dodecatungstophosphoric acid (DTP)/K-10 montomorillonite clay.
  • Synthesize 3,5-dimethylbenzaldehyde from benzene via peroxidative oxidation in the presence of vanadium(IV or V) compounds with N,O-ligands as catalysts.

Packaging

10L packaged in barrier layer poly jerrycan with electrostatically dissipating outside; 5mL, 100mL, 500mL packaged in glass bottles

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

122.0 °F

Flash Point(C)

50 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peroxidative oxidation of benzene and mesitylene by vanadium catalysts.
Reis PM, et al.
J. Mol. Catal. A: Chem., 224(1), 189-195 (2004)
Carbon-13 Relaxation and Internal Rotation in Mesitylene and o-Xylene.
Kuhlmann KF and Grant DM.
J. Chem. Phys. , 55(6), 2998-3007 (1971)
The molecular structure of mesitylene as determined by electron diffraction.
Almenningen, A, et al.
Journal of Molecular Structure, 96(3-4), 373-377 (1983)
Growth mechanism of C60/mesitylene nanowires.
Zhou Y and Zhou W.
CrystEngComm, 14(4), 1449-1454 (2012)
Kinetics and mechanism of selective monoacylation of mesitylene.
Yadav GD and Asthana NS.
Industrial & Engineering Chemistry Research, 41(23), 5565-5575 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service