Skip to Content
Merck
All Photos(1)

Key Documents

T73601

Sigma-Aldrich

1,2,4-Trimethylbenzene

98%

Synonym(s):

Pseudocumene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3(CH3)3
CAS Number:
Molecular Weight:
120.19
Beilstein:
1903005
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39011403
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.1 (vs air)

vapor pressure

4.5 mmHg ( 37.7 °C)

Assay

98%

autoignition temp.

959 °F

expl. lim.

6.4 %

refractive index

n20/D 1.504 (lit.)

bp

168 °C (lit.)

mp

−44 °C (lit.)

density

0.876 g/mL at 20 °C (lit.)

SMILES string

Cc1ccc(C)c(C)c1

InChI

1S/C9H12/c1-7-4-5-8(2)9(3)6-7/h4-6H,1-3H3

InChI key

GWHJZXXIDMPWGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2,4-Trimethylbenzene, also known as Pseudocumene is an alkylated aromatics that is commonly used in the synthesis of polycyclic aromatic hydrocarbons.

Application

1,2,4-Trimethylbenzene can be used as a reactant to synthesize:
  • 2,3,5-Trimethylbenzoquinone via oxidation reaction with phthaloyl peroxide under solvent-free conditions.      
  • 1,2,4,5-Tetramethylbenzene (durene) by methylation using syngas in the presence of Cu and Zn-based bifunctional catalysts.
  •  1,2,4-Oxadiazole derivatives via tandem reaction with nitroalkenes and nitriles in the presence of superacid CF3SO3H (TfOH).

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

118.4 °F - closed cup

Flash Point(C)

48.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of durene by methylation of 1, 2, 4-trimethylbenzene with syngas over bifunctional CuZnZrO x-HZSM-5 catalysts
Wen D, et al.
Catalysis Science & Technology (2022)
Polycyclic aromatic hydrocarbon formation in a flame of the alkylated aromatic trimethylbenzene compared to those of the alkane dodecane
Tirthankar M, et al.
Combustion and Flame, 223, 495-510 (2021)
An efficient synthesis of 2, 3, 5-trimethylbenzoquinone by metal-free oxidation of 1, 2, 4-trimethylbenzene
Yerramreddy TR, et al.
J. Chem. Res. (M), 43(11-12), 565-568 (2019)
Andrei A Golushko et al.
The Journal of organic chemistry, 84(11), 7495-7500 (2019-05-24)
The tandem reaction of nitroalkenes with arenes and nitriles in the superacid CF3SO3H (TfOH) results in the formation of 1,2,4-oxadiazole derivatives in yields up to 96%. This reaction proceeds via the consequent interaction of arenes and nitriles, as nucleophiles, with
C M Kao et al.
Water research, 35(8), 1951-1960 (2001-05-08)
Natural bioremediation is believed to be the major processes that account for both containment of the petroleum-hydrocarbon plume and reduction of the contaminant concentrations. In this study, the feasibility of applying the microbial enumeration technique was assessed for natural biodegradation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service