Skip to Content
Merck
All Photos(2)

Key Documents

484431

Sigma-Aldrich

Propylene glycol monomethyl ether acetate

ReagentPlus®, ≥99.5%

Synonym(s):

DOWANOL® PMA, 1,2-Propanediol monomethyl ether acetate, 1-Methoxy-2-propyl acetate, MPA, PGMEA, Propylene glycol methyl ether acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH(CH3)CH2OCH3
CAS Number:
Molecular Weight:
132.16
Beilstein:
1751656
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
bp:
145-146 °C (lit.)
145-146 °C
vapor pressure:
3.7 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

reagent

Quality Level

vapor pressure

3.7 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

≥99.5%

autoignition temp.

669 °F

expl. lim.

13.1 %

dilution

(for general lab use)

refractive index

n20/D 1.402 (lit.)

bp

145-146 °C (lit.)
145-146 °C

solubility

water: soluble 198 g/L at 20 °C

density

0.970 g/mL at 25 °C (lit.)

functional group

ester
ether

SMILES string

COCC(C)OC(C)=O

InChI

1S/C6H12O3/c1-5(4-8-3)9-6(2)7/h5H,4H2,1-3H3

InChI key

LLHKCFNBLRBOGN-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Propylene glycol monomethyl ether acetate (PGMEA) is a photoresist solvent.[1] Its degradation by microorganisms in different soil types has been investigated.[2] An oral reference dose (RfD) value of PGMEA has been obtained from inhalation studies.[3] The solubility of (5-alkylsulfonyloxyimino-5H-thiophen-2-ylidene)-2-methylphenyl-acetonitriles in PGMEA has been analyzed.[4]

Application

Propylene glycol monomethyl ether acetate has been used in the development of fluidic channels to be incorporated in a poly(dimethylsiloxane) (PDMS)-based microfluidic device.[5]

Legal Information

Product of Dow Chemical
ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
DOWANOL is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

113.9 °F - closed cup

Flash Point(C)

45.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian Bleilevens et al.
Scientific reports, 8(1), 8031-8031 (2018-05-26)
Extracorporeal lung assist technology is one of the last options in critical care medicine to treat patients suffering from severe oxygenation and decarboxylation disorders. Platelet activation along with the consequent thrombus formation is a potentially life-threatening complication of this technique.
Novel photoacid generators for chemically amplified resists with g-line, i-line, and DUV exposure.
Asakura T, et al.
SPIE Proc., 4345 (2001)
Yug C Saraswat et al.
Journal of colloid and interface science, 564, 43-51 (2020-01-07)
Our ability to dictate the colloid geometry is intimately related to self-assembly. The synthesis of anisotropic colloidal particles is currently dominated by wet chemistry and lithographic techniques. The wet chemical synthesis offers limited particle geometries at bulk quantities. Lithographic techniques
Comparison of photoresist shelf life in PGMEA and CA solvent systems.
Nelson WC.
SPIE Proc., 2195 (1994)
Nasim Annabi et al.
Lab on a chip, 13(18), 3569-3577 (2013-06-04)
The research areas of tissue engineering and drug development have displayed increased interest in organ-on-a-chip studies, in which physiologically or pathologically relevant tissues can be engineered to test pharmaceutical candidates. Microfluidic technologies enable the control of the cellular microenvironment for

Questions

1–3 of 3 Questions  
  1. What is the makeup for this fluid? Does it have any harmful contents for humans?

    1 answer
    1. This product is Propylene glycol monomethyl ether acetate. It is intended for use in a chemistry lab setting and should not be used on human or animal subjects.

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service