Skip to Content
Merck
All Photos(3)

Key Documents

296333

Sigma-Aldrich

p-Xylene

anhydrous, ≥99%

Synonym(s):

1,4-Dimethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH3)2
CAS Number:
Molecular Weight:
106.17
Beilstein:
1901563
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
Assay:
≥99%
bp:
138 °C (lit.)
vapor pressure:
9 mmHg ( 20 °C)

grade

anhydrous

Quality Level

vapor density

3.7 (vs air)

vapor pressure

9 mmHg ( 20 °C)

Assay

≥99%

form

liquid

autoignition temp.

984 °F

expl. lim.

7 %

impurities

<0.002% water
<0.005% water (100 mL pkg)

evapn. residue

<0.0003%

refractive index

n20/D 1.495 (lit.)

bp

138 °C (lit.)

mp

12-13 °C (lit.)

solubility

water: soluble 0.2 g/L

density

0.861 g/mL at 20 °C (lit.)

SMILES string

Cc1ccc(C)cc1

InChI

1S/C8H10/c1-7-3-5-8(2)6-4-7/h3-6H,1-2H3

InChI key

URLKBWYHVLBVBO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

p-Xylene is an isomer xylene mainly used in the polyester industry as a raw material for synthesizing 5,6-terephthalic acid.

Application

p-Xylene may be used in the synthesis of ethyl 2,5-dimethylphenylhydroxymalonate by reacting with ethyl oxomalonate in the presence of anhydrous stannic chloride.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

2,5-dimethylmandelic acid
Riebsomer JL and Irvine J
Organic Syntheses, 25, 33-33 (1945)
Joshua B Marlow et al.
Physical chemistry chemical physics : PCCP, 20(24), 16592-16603 (2018-06-07)
Lamellar liquid crystals comprising oil, water and surfactant(s) were formulated and analysed in order to examine how these materials responded to the inclusion of inorganic nanoparticles, in terms of their structural and rheological characteristics. Lamellar phases were formed from mixtures
Friedel-Crafts acylation of toluene and p-xylene with carboxylic acids catalyzed by zeolites.
Chiche B, et al.
The Journal of Organic Chemistry, 51(11), 2128-2130 (1986)
Cycloaddition of biomass-derived furans for catalytic production of renewable p-xylene.
Williams CL, et al.
ACS Catalysis, 2(6), 935-939 (2012)
Ultra-selective cycloaddition of dimethylfuran for renewable p-xylene with H-BEA.
Chang CC, et al.
Green Chemistry, 16(2), 585-588 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service