Skip to Content
Merck
All Photos(1)

Documents

B71608

Sigma-Aldrich

2-Bromomesitylene

98%

Synonym(s):

2-Bromo-1,3,5-trimethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
1,3,5-(CH3)3C6H2Br
CAS Number:
Molecular Weight:
199.09
Beilstein:
1907245
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.552 (lit.)

bp

225 °C (lit.)

mp

2 °C (lit.)

density

1.301 g/mL at 25 °C (lit.)

SMILES string

Cc1cc(C)c(Br)c(C)c1

InChI

1S/C9H11Br/c1-6-4-7(2)9(10)8(3)5-6/h4-5H,1-3H3

InChI key

RRTLQRYOJOSPEA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Bromomesitylene can be used as a reactant for carbon-carbon bond formation and cleavage reactions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nitrile-group transfer from solvents to aryl halides. novel carbon- carbon bond formation and cleavage mediated by palladium and zinc species
Luo FH, et al.
Organometallics, 17, 1025-1030 (1998)
Karlee L Bamford et al.
Dalton transactions (Cambridge, England : 2003), 49(48), 17571-17577 (2020-11-25)
The synthesis and isolation of the first stable C-B-N-substituted borinium [MesBNiPr2][B(C6F5)4] (2) is described. Compound 2 was shown to react with isothiocyanate and carbodiimides, effecting B-C insertion to afford nitrilium (4) and borenium amidinate salts, respectively. The borinium cation [MesBNiPr2]+

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service