Skip to Content
Merck
All Photos(1)

Key Documents

222461

Sigma-Aldrich

Murexide

ACS reagent

Synonym(s):

5,5′-Nitrilodibarbituric acid monoammonium salt, Ammonium purpurate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8N6O6
CAS Number:
Molecular Weight:
284.19
Colour Index Number:
56085
Beilstein:
3831445
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

ACS reagent

Quality Level

form

powder

technique(s)

titration: suitable

mp

>300 °C (lit.)

λmax

525 nm

suitability

passes test for complexometric indicator

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

N.OC1=C(\N=C2\C(=O)NC(=O)NC2=O)C(=O)NC(=O)N1

InChI

1S/C8H5N5O6.H3N/c14-3-1(4(15)11-7(18)10-3)9-2-5(16)12-8(19)13-6(2)17;/h(H2,10,11,14,15,18)(H3,12,13,16,17,19);1H3

InChI key

LJYRLGOJYKPILZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Murexide is the ammonium salt of purpuric acid that appears as a slightly reddish-purple powder and is soluble in water. It is mainly used as an indicator for complexometric titrations to measure water hardness and as a colorant/pigment in the dye industry.

Application

Murexide is used for the histochemical staining of calcium in the skeleton. It is suitable for staining the hemijaws for dental caries scoring.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Ghaedi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 83(1), 46-51 (2011-09-10)
In the first, Cadmium selenide Nanoparticle loaded on activated carbon (CdSe-NP-AC) has been synthesized and characterized by different techniques including XRD and SEM. Then, this new adsorbent successfully has been applied for the removal of muroxide (MO) from aqueous solution
A Hirota et al.
The Journal of general physiology, 94(4), 597-631 (1989-10-01)
Two new Ca indicators, purpurate-3,3'diacetic acid (PDAA) and 1,1'-dimethylpurpurate-3,3'diacetic acid (DMPDAA), were synthesized and used to measure Ca transients in frog cut muscle fibers. These indicators are analogues of the purpurate components of murexide and tetramethylmurexide, in which two acetate
F D Coffman et al.
Biochemistry, 27(16), 6179-6187 (1988-08-09)
An insulin hexamer containing one B10-bound Co(III) ion and one unoccupied B10 site has been synthesized. The properties of the monosubstituted hexamer show that occupancy of only one B10 site by Co3+ is sufficient to stabilize the hexameric form under
Putting an old dye to a new use.
Gonzalo Pizarro
The Journal of physiology, 581(Pt 1), 4-4 (2007-03-24)
R De Giovanni et al.
Physiological chemistry and physics and medical NMR, 24(4), 271-280 (1992-01-01)
In this paper the authors studied the effects of thyroid hormones and their structural analogues on the mitochondrial calcium transport activities. The thyroid hormones, 3,5,3' L-triiodothyronine (LT3) and 3,5,3'5' L-tetraiodothyronine (LT4) at physiological intracellular concentrations between 7.2 and 9 nM

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service