Skip to Content
Merck
All Photos(4)

Documents

114510

Sigma-Aldrich

Methyl Orange

ACS reagent, Dye content 85 %

Synonym(s):

4-[4-(Dimethylamino)phenylazo]benzenesulfonic acid sodium salt, Acid Orange 52, Helianthin, Orange III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14N3NaO3S
CAS Number:
Molecular Weight:
327.33
Colour Index Number:
13025
Beilstein:
4732884
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

ACS reagent

form

powder

composition

Dye content, 85%

technique(s)

titration: suitable

color

orange

pH

3.0-4.4, pink to yellow

visual transition interval

3.2-4.4, red to yellow

mp

>300 °C

λmax

505 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].CN(C)c1ccc(cc1)\N=N\c2ccc(cc2)S([O-])(=O)=O

InChI

1S/C14H15N3O3S.Na/c1-17(2)13-7-3-11(4-8-13)15-16-12-5-9-14(10-6-12)21(18,19)20;/h3-10H,1-2H3,(H,18,19,20);/q;+1/p-1/b16-15+;

InChI key

STZCRXQWRGQSJD-GEEYTBSJSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Methyl orange (C.I. 13025) is the sodium salt of p-dimethylaminoazobenzenesulfonic acid.It is an orange-colored synthetic dye, which changes color depending on the pH of the solution. It develops a red color in acidic solution and yellow color in basic solution.

Application

Methyl orange is used as a-
  • pH indicator in the cell sap
  • counterstain to crystal violet in staining pollen tubes
  • component of a polychrome histological stain
  • solution indicator, such as in assays of oxalic and malonic acids
  • in other biomedical applications that include use in film dosimeters and as a reagent for the assay of bromide ions.

Biochem/physiol Actions

Methyl Orange is an azo dye which works as a pH indicator.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Encapsulation of methyl orange pH-indicator into a sol-gel matrix.
Zaggout FR, et al.
Materials Letters, 59, 2928-2931 (2005)
Somayeh Mirsadeghi et al.
Journal of environmental management, 270, 110831-110831 (2020-06-09)
A simple biogenic approach for synthesis of Fe3O4/Au nanocomposite with 31 nm size using aqueous extract of Carum carvi L. seeds has been reported. Phytochemicals of Carum carvi L. seeds extract play three roles, including reducing, capping, and stabilizing agents during
Characterization of methyl orange and its photocatalytic degradation products by HPLC/UV?VIS diode array and atmospheric pressure ionization quadrupole ion trap mass spectrometry.
Baiocchi C, et al.
International Journal of Mass Spectrometry, 214, 247-256 (2002)
Soon-An Ong et al.
Environmental science and pollution research international, 20(5), 3405-3413 (2012-11-02)
The objective of this study was to examine the effects of adsorbability and number of sulfonate group on solar photocatalytic degradation of mono azo methyl orange (MO) and diazo Reactive Green 19 (RG19) in single and binary dye solutions. The
Jie Li et al.
Nanotechnology, 24(15), 155603-155603 (2013-03-23)
We report on the synthesis of high-quality microporous/mesoporous BN material via a facile two-step approach. An extremely high surface area of 1687 m(2) g(-1) and a large pore volume of 0.99 cm(3) g(-1) have been observed in the synthesized BN porous whiskers. The formation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service