Skip to Content
Merck
All Photos(1)

Key Documents

117706

Sigma-Aldrich

Phenylacetylene

98%

Synonym(s):

Ethynylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CCH
CAS Number:
Molecular Weight:
102.13
Beilstein:
605461
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

17.6 mmHg ( 37.7 °C)

Quality Level

Assay

98%

impurities

<1% 1,4-dioxane

refractive index

n20/D 1.549 (lit.)

bp

142-144 °C (lit.)

solubility

H2O: insoluble
alcohol: miscible
diethyl ether: miscible

density

0.93 g/mL at 25 °C (lit.)

functional group

phenyl

storage temp.

2-8°C

SMILES string

C#Cc1ccccc1

InChI

1S/C8H6/c1-2-8-6-4-3-5-7-8/h1,3-7H

InChI key

UEXCJVNBTNXOEH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phenylacetylene is an aromatic organic compound frequently employed in Sonogashira coupling reactions. Phenylacetylene undergoes polymerization catalyzed by Rh and Pt complexes to form polyphenylacetylene.

Application

Phenylacetylene was used in a study to investigate the mechanism of product formation during the palladium-catalysed phenylacetylene oxidative carbonylation reaction.
Terminal acetylene used in the conversion of nitrones to alkynyl hydroxyl amines in the presence of trimethylaluminum..

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rhodium and platinum complexes as catalysts for the polymerization of phenylacetylene.
Furlani A, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 24(5), 991-1005 (1986)
Tetrahedron Letters, 48, 1457-1457 (2007)
Katarina Novakovic et al.
Physical chemistry chemical physics : PCCP, 10(5), 749-753 (2008-02-07)
This paper reports on the influence of oscillations on product selectivity as well as the dynamics of product formation during the palladium-catalysed phenylacetylene oxidative carbonylation reaction in a catalytic system (PdI2, KI, Air, NaOAc in methanol). The occurrence of the
Zhouqing Xu et al.
Chemical communications (Cambridge, England), 48(46), 5736-5738 (2012-05-09)
[8+12]-metallamacrocycle-based 3D frameworks {[Cu(4)(pbt)(2)(SO(4))(2)(DMF)(2)(CH(3)OH)]·7H(2)O·DMF}(n) (1) and [12]-macrocycle 3D {[Cu(2)(pbt)(SO(4))(DMSO)(CH(3)OH)(2)]·5H(2)O·CH(3)OH}(n) (2) have been obtained. Both complexes display antiferromagnetic couplings and high catalytic activity in the oxidative coupling reaction of 1-ethynylbenzene and oxazolidin-2-one.
Zhuofeng Ke et al.
Journal of the American Chemical Society, 134(37), 15418-15429 (2012-09-13)
Artificial metalloenzyme, composed of metal complex(es) and a host protein, is a promising way to mimic enzyme catalytic functions or develop novel enzyme-like catalysis. However, it is highly challenging to unveil the active site and exact reaction mechanism inside artificial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service