Skip to Content
Merck
All Photos(1)

Key Documents

Y0000754

Methenamine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Hexamethylenetetramine, 1,3,5,7-Tetraazatricyclo[3.3.1.13,7]decane, Hexamine, Methenamine, Urotropine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12N4
CAS Number:
Molecular Weight:
140.19
Beilstein:
2018
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor pressure

<0.01 mmHg ( 20 °C)

API family

methenamine

autoignition temp.

770 °F

manufacturer/tradename

EDQM

mp

280 °C (subl.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

C1N2CN3CN1CN(C2)C3

InChI

1S/C6H12N4/c1-7-2-9-4-8(1)5-10(3-7)6-9/h1-6H2

InChI key

VKYKSIONXSXAKP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Methenamine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2 - Skin Sens. 1B

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

482.0 °F - closed cup

Flash Point(C)

250 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adriana Luk et al.
The Malaysian journal of pathology, 36(2), 71-81 (2014-09-10)
The incidence of infective endocarditis is 1.5-4.95 cases per 100,000 individuals per year, with a mortality of 14-46% 1-year post infection. The management and decision to operate on selected patients remains controversial. Our study reviews cases of native and prosthetic
Mehdi Taghdiri et al.
Journal of hazardous materials, 246-247, 206-212 (2013-01-15)
The industrial wastewater produced by hexamine plants is considered as a major environmental polluting factor due to resistance to biodegradation. So the treatment of such wastewater is required. In this work, the removal of hexamine from wastewater and its degradation
Jianguo Hu et al.
Talanta, 93, 345-349 (2012-04-10)
A green, one-step method for synthesis of graphene-Au nanoparticles (graphene-AuNPs) was introduced in this article, using an environmentally benign hexamethylenetetramine (HMTA) as reducing and stabilizing agent. HMTA slowly was hydrolyzed to generate aldehyde ammonia to reduce graphene oxides (GO) and
Ann Pontén et al.
Contact dermatitis, 68(1), 50-53 (2012-10-06)
The currently used patch test concentration for formaldehyde is 1.0% (wt/vol) in water. However, clinical experience and previous studies suggest that 1.0% might be insufficient for detecting an optimized number of clinically relevant cases of contact allergy to formaldehyde. To
Lorena Vidal et al.
Journal of chromatography. A, 1285, 7-14 (2013-03-12)
Quaternary ammonium-functionalized silica materials were synthesized and applied for solid-phase extraction (SPE) of aromatic amines, which are classified as priority pollutants by US Environmental Protection Agency. Hexamethylenetetramine used for silica surface modification for the first time was employed as SPE

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service